Ergo-Tweeze® Fine Point Tweezers, Excelta®
Catalog # 75880-780
Supplier: Excelta
Specifications
- Material:Stainless steel, anti-magnetic
- Cat. no.:75880-780
- Overall Length:121 mm (4.75")
- Supplier No.:3C-SA-SE-ET
Specifications
About this item
These very fine pointed tweezers are ideal for applications requiring high magnification.
- Ergo-Tweeze® soft cushioned grips
- Ideal for handling ESD sensitive components (10¹⁰ ohms/sq.)
Ideal for repetitive handling tasks in specimen preparation, electronics, instrumentation, laboratories, and forensics.