Replacement Tweezer Tips, Excelta Corp®
Replacement Tweezer Tips, Excelta Corp®
 :   Excelta
Replacement Tweezer Tips, Excelta Corp®
 :  Excelta
Replacement Tweezer Tips, Excelta Corp®
Replacement Tweezer Tips, Excelta Corp®
Replacement tips for tweezer model 162D-RT. Peek is a high temperature thermoplastic with excellent chemical resistance.
Including screws.
Product Family Options
  • Désignation
    Matière
    Tip Length
    Tip Width
    Curvature
  • Replacement tips for 162d-rt, 5 star quality
    PEEK
    0.4 mm (0.016")
    0.08" (2 mm)
    Straight
    Each
    Each - $87.61
    $87.61
    Each
       $87.61
     : 162D-RTX
  • Replacement tips for 162b-rt, 5 star quality
    PEEK
    0.7 mm (0.03")
    0.03" (0.7 mm)
    Straight
    Each
    Each - $124.93
    $124.93
    Each
       $124.93
     : 162B-RTX
  • Replacement tips for 162a-rt, 5 star quality
    PEEK
    0.5 mm (0.02")
    0.02" (0.5 mm)
    Straight
    Each
    Each - $122.33
    $122.33
    Each
       $122.33
     : 162A-RTX
  • Replacement tips for 762a-rt, 5 star quality
    PEEK
    0.3 mm (0.012")
    0.02" (0.5 mm)
    50°
    Each
    Each - $95.12
    $95.12
    Each
       $95.12
     : 762A-RTX