VWR® LTS Compatible Pipette Tips
About this item
VWR® LTS™ Compatible pipette tips feature a flexible seal area and a positive stop resulting in a secure seal with light ejection force on LTS™ compatible pipettors.
- Compatible with the LTS™ style pipettors
- Maximizes sample recovery with low binding resin and a fine point design
- Improves ergonomics by reducing the force required to attach and eject a tip
- Autoclavable at 15 psi, 122 °C for 15 minutes
- Manufactured under an ISO 9001 quality management system
VWR® tips use a low retention resin that is up to ten times more hydrophobic than other polypropylene pipette tips, enabling viscous samples like protein and DNA solutions to dispense easily without collecting inside the tip.
Certifications: Certified RNase-, DNase-, endotoxin-, metal-, and human DNA-free.
25 Options Available Below
Product Details & Documents
VWR® LTS™ Compatible pipette tips feature a flexible seal area and a positive stop resulting in a secure seal with light ejection force on LTS™ compatible pipettors.
- Compatible with the LTS™ style pipettors
- Maximizes sample recovery with low binding resin and a fine point design
- Improves ergonomics by reducing the force required to attach and eject a tip
- Autoclavable at 15 psi, 122 °C for 15 minutes
- Manufactured under an ISO 9001 quality management system
VWR® tips use a low retention resin that is up to ten times more hydrophobic than other polypropylene pipette tips, enabling viscous samples like protein and DNA solutions to dispense easily without collecting inside the tip.
Certifications: Certified RNase-, DNase-, endotoxin-, metal-, and human DNA-free.
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
- Catalog No:
- 76322-158
- 76322-160
- 76322-162
- 76322-164
- 76327-216
- 76323-944
- 76323-946
- 76323-932
- 76323-934
- 76327-218
- 76323-936
- 76323-938
- 76323-940
- 76323-942
- 76327-220
- 76322-530
- 76322-532
- 76322-534
- 76322-180
- 76327-212
- 76322-172
- 76322-174
- 76322-176
- 76322-178
- 76327-210
- Environmentally Preferable:
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Waste Reducing|Sustainable Packaging
- Environmental certification:
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%
- FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%
Specifications
Product Family Options
Racked, filtered, sterile
Product Information
- VolumeSterileLongueurPackedAvailabilityPrice
- 0.1 - 20 µl+49.63 mm (1.95")10×96 Tips/PackPack of 960$198.49Case of 4800$992.46Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76322-158Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging - 200 µl+50.75 mm (2.00")10×96 Tips/PackPack of 960$196.40Case of 4800$982.01Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76322-160Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging - 20 - 300 µl+58.93 mm (2.32")10×96 Tips/PackPack of 960$185.50Case of 4800$927.51Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76322-162Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging - 100 - 1000 µl+88.9 mm (3.50")8×96 Tips/PackPack of 768$199.19Case of 3840$995.96Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76322-164Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging - 100 - 1200 µl+97.66 mm (3.85")8×96 Tips/PackPack of 768$189.38Case of 3840$946.90Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76327-216Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging
Racked, non-filtered, sterile
Product Information
- VolumeSterileLongueurPackedAvailabilityPrice
- 0.1 - 20 µl+49.63 mm (1.95")10×96 Tips/PackPack of 960$111.27Case of 4800$556.35Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76323-944Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging - 250 µl+50.75 mm (2.00")10×96 Tips/PackPack of 960$111.93Case of 4800$559.64Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76323-946Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging - 20 - 300 µl+58.93 mm (2.32")10×96 Tips/PackPack of 960$85.76Case of 4800$428.81Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76323-932Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging - 100 - 1000 µl+88.9 mm (3.50")8×96 Tips/PackPack of 768$111.62Case of 3840$558.12Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76323-934Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging - 100 - 1200 µl+97.66 mm (3.85")8×96 Tips/PackPack of 768$88.20Case of 3840$441.00Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76327-218Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging
Racked, non-filtered, non-sterile
Product Information
- VolumeSterileLongueurPackedAvailabilityPrice
- 0.1 - 20 µl49.63 mm (1.95")10×96 Tips/PackPack of 960$88.35Case of 4800$441.77Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76323-936Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging - 250 µl50.75 mm (2.00")10×96 Tips/PackPack of 960$92.89Case of 4800$464.47Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76323-938Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging - 20 - 300 µl58.93 mm (2.32")10×96 Tips/PackPack of 960$93.22Case of 4800$466.08Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76323-940Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging - 100 - 1000 µl88.9 mm (3.50")8×96 Tips/PackPack of 768$91.96Case of 3840$459.82Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76323-942Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging - 100 - 1200 µl97.66 mm (3.85")8×96 Tips/PackPack of 768$81.96Case of 3840$409.78Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76327-220Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plasticWaste Reducing|Sustainable Packaging
Reload, non-filtered, sterile
Product Information
- VolumeSterileLongueurPackedAvailabilityPrice
- 0.1 - 20 µl+49.63 mm (1.95")10×96 Tips/PackPack of 960$74.16Case of 4800$370.79Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76322-530Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%Waste Reducing|Sustainable Packaging - 250 µl+50.75 mm (2.00")10×96 Tips/PackPack of 960$89.45Case of 4800$447.24Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76322-532Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%Waste Reducing|Sustainable Packaging - 20 - 300 µl+58.93 mm (2.32")10×96 Tips/PackPack of 960$85.97Case of 4800$429.83Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76322-534Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%Waste Reducing|Sustainable Packaging - 100 - 1000 µl+88.9 mm (3.50")8×96 Tips/PackPack of 768$85.06Case of 3840$425.28Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76322-180Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%Waste Reducing|Sustainable Packaging - 100 - 1200 µl+97.66 mm (3.85")8×96 Tips/PackPack of 768$87.50Case of 3840$437.48Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76327-212Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%Waste Reducing|Sustainable Packaging
Reload, non-filtered, non-sterile
Product Information
- VolumeSterileLongueurPackedAvailabilityPrice
- 0.1 - 20 µl49.63 mm (1.95")10×96 Tips/PackPack of 960$62.30Case of 4800$311.51Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76322-172Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%Waste Reducing|Sustainable Packaging - 250 µl50.75 mm (2.00")10×96 Tips/PackPack of 960$74.60Case of 4800$372.98Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76322-174Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%Waste Reducing|Sustainable Packaging - 20 - 300 µl58.93 mm (2.32")10×96 Tips/PackPack of 960$70.07Case of 4800$350.36Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76322-176Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%Waste Reducing|Sustainable Packaging - 100 - 1000 µl88.9 mm (3.50")8×96 Tips/PackPack of 768$68.86Case of 3840$344.29Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76322-178Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%Waste Reducing|Sustainable Packaging - 100 - 1200 µl97.66 mm (3.85")8×96 Tips/PackPack of 768$56.28Case of 3840$281.41Specifications:
Waste Reducing|Sustainable Packaging More
Specifications
Cat. No.76327-210Environmentally PreferableWaste Reducing|Sustainable PackagingEnvironmental certificationFSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49%Waste Reducing|Sustainable Packaging
Empty racks
Product Information
- Catalog #For Use WithDésignationAvailabilityPrice
- 1000 - 1250 µl tipsEmpty Racks for Reloads, Autoclavable, 10 Racks/PackLog in for availabilityPack of 10$92.07Case of 40$92.07