Seismograph Model
Seismograph model
Catalog # 470324-782
Supplier: Wards
: BG0014
Jump to:
Product Details & Documents
Clearly demonstrate how a seismograph records the shock waves generated by an earthquake in the classroom or laboratory.
- Heavy duty, premium seismograph model
- Can be used to study both geological and mathematical phenomena
- Writing instrument is not included
This seismograph model creates a visual representation of the shock waves generated by an earthquake. It can be used to teach both mathematical and geological concepts in an educational setting. Demonstration is conducted by manually pulling the paper strip and recording simple movements made by jumping or moving next to the model. This model is designed for educational concept demonstration and is not intended to be used to accurately measure seismic waves.
Specifications
- Désignation:Seismograph model