Celestial Star Globe | Avantor
Celestial Star Globe
12" Celestial star globe
Catalog # 470324-780
Supplier:   Wards
Fournisseur #:  BD0077CSG
Celestial Star Globe
Catalog # 470324-780
Supplier:  Wards
Celestial Star Globe
Celestial Star Globe
12" Celestial star globe
Price
$159.35
Log in pour la disponibilité
Each
Product Details & Documents
Perfect for introductory-level astronomy students.
  • 12" Globe diameter
  • 17.5" Tall (overall model)
  • A 3" detailed earth globe is fitted inside the large star globe
This stunning star globe illustrates the effects of Earth’s rotation on its axis and its revolution around the sun in addition to depicting the constellations, Milky Way, and major stars in three dimensions. Color-coding and clear silk-screened names make it easy to identify the many stars, while celestial latitude and longitude scales provide orientation. A detailed 3" Earth globe is centered in the larger star globe. Both globes can be set to the positions of the stars and planets for any time and place on Earth in the past, present, or future.
Spécifications
  • Désignation:
    12" Celestial star globe