Bags, Cell Culture, Ultra Clear, 6-Port, Flex Concepts, Chemglass
Fournisseur: Chemglass
Certificats
À propos de cet article
Bags are compatible with GE Wave™ rocking platforms.
- Contamination free
Cell culture bags are manufactured from ultra clear Flex 600 robust film and gamma irradiated. The inner product contact material is low density polyethylene. Excellent choice for mammalian and plant culture cell lines with extended mixing times.
Certifications: Meets USP Class VI specifications.
Frequently Bought Together