ContactUsLinkComponent
Gasket, Perfluoro, Black, 400 mm DBR Industries
Gasket, Perfluoro, Black, 400 mm DBR Industries
Catalog # 76333-492
Supplier:  Chemglass
undefined
Gasket, Perfluoro, Black, 400 mm DBR Industries
Catalog # 76333-492
Supplier:  Chemglass
Supplier Number:  CG-1968-H-02
Specifications
  • Description:
    Gasket, 400 mm, perfluoroelastomer, centering tabs, class vi ffkm, -15...275 °c
  • Cat. no.:
    76333-492
Specifications
About this item
Class VI Perfluoroelastomer with 3 tabs each spaced 120 degrees apart along the outer edge to aid in gasket alignment.
  • Gasket is for use with 400 mm Chemglass flat ground flange on Chemglass 50 and 100 L vessels
The chemically resistant perfluoroelestomer has a soft pliable sealing surface that provides improved sealing under full vacuum, as opposed to encapsulated o-ring materials and envelope gaskets.