Electronic Cell Stirrer, FireflySci | Avantor
Electronic Cell Stirrer, FireflySci
Marketsource
Supplier:   FIREFLYSCI, INC.
Electronic Cell Stirrer, FireflySci
Supplier:  FIREFLYSCI, INC.
Marketsource
Certificates
Electronic Cell Stirrer, FireflySci
Electronic Cell Stirrer, FireflySci
2 Options Available Below
Product Details & Documents
Stirs cell contents while measuring absorbance or fluorescence. Fits all standard spectrophotometers and fluorometers with a 10 mm cuvette holder.
  • Controller and electronic magnetic stirring assembly included
  • Controller is 4.0×2.5×3.0" (including speed controller knob)
  • Available for use with 100/120 or 220/240 VAC wall currents
The sample is mixed by a tiny magnetic stir bar which is placed at the bottom of the cell. A small electronic rotation magnet assembly is placed under the cell in the cell compartment. This raises the cell up 5mm, which allows use of less sample volume, and will not interfere with the unit's operation.
Mixer rotation speed is controlled by an external controller, connected by a 36" long paper thin ribbon, allowing the instrument to close normally.
Ordering information: Each electronic cell stirrer comes with a P73 Magnetic stir bar that fits any 10×10 mm cell, and will stir up to 30 mm in internal height.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Electronic cell Stirrer, 110V
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,455.25
    $4,455.25
    Each
       $4,455.25
    La quantité minimale de commande est de 1
    Supplier Number: P82-110VAC
    Spécifications:
    Marketsource
    Marketsource
  • Electronic cell Stirrer, 240V
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,455.25
    $4,455.25
    Each
       $4,455.25
    La quantité minimale de commande est de 1
    Supplier Number: P82-240VAC
    Spécifications:
    Marketsource
    Marketsource