Vernier® Human Physiology Experiments Lab Book
Human physiology experiments lab book
CA470320-006
New Product
: HSB-HP
Vernier® Human Physiology Experiments Lab Book
CA470320-006
New Product
Ready-to-use student experiments.
- Electronic access to up-to-date versions of the experiments
- Instructions for data collection with Graphical Analysis 4 app with Go Direct sensors. (Does not support data collection with Logger Pro or LabQuest App)
- Essential instructor information, including teaching tips, suggested answers, and sample data and graphs
- Word-processing files of the student experiments, so you can edit the files to match your teaching preferences
- Complete equipment and supplies list
This contains 14 experiments designed to encourage students to explore the physiology of various human organ systems. This lab book provides instructions for data collection with Graphical Analysis™ 4 and Go Direct® sensors only.
- Désignation:Human physiology experiments lab book