Micro Drop Spectrophotometer Conversion Kit, Micro Focus Cell (1.9 µl), FireflySci | Avantor
Micro Drop Spectrophotometer Conversion Kit, Micro Focus Cell (1.9 µl), FireflySci
Micro drop spectrophotometer conversion kit, 1.9 µl
Catalog # 76016-870
Marketsource
Supplier:   FIREFLYSCI, INC.
Fournisseur #:  MFC-1
Micro Drop Spectrophotometer Conversion Kit, Micro Focus Cell (1.9 µl), FireflySci
Catalog # 76016-870
Supplier:  FIREFLYSCI, INC.
Marketsource
Micro Drop Spectrophotometer Conversion Kit, Micro Focus Cell (1.9 µl), FireflySci
Micro Drop Spectrophotometer Conversion Kit, Micro Focus Cell (1.9 µl), FireflySci
Micro drop spectrophotometer conversion kit, 1.9 µl
Price
$12,134.75
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Transform a current spectrophotometer into a nano-sized spectrophotometer with the Micro focus cell conversion kit.
  • Ultra-micro low volume cell with a 1.9 µl capacity
  • Convert any spectrophotometer into a high end drop based machine
  • Leak-proof design
Applications include DNA measurement, RNA measurement, DNA quantification, nucleic acid analysis, protein concentration, and all small volume applications (0.3 to 1.9 µl). Quickly load and empty a sample into the cell without even removing it from the machine. This results in consistent alignment through multiple tests.
Ordering information: Each kit includes a 12.5×12.5 micro focus adapter that fits into any spectrophotometer, a UV Quartz micro tray to inject a sample into, spacers to adjust the micro focus adapter to the proper Z-Dimension in the spectrophotometer, cleaning swabs, and instructions.
Spécifications
  • Capacité:
    1.9 µl
  • Désignation:
    Micro drop spectrophotometer conversion kit