Laboratory Warning Labels
À propos de cet article
Various warning and safety labels for use throughout the lab. Designed to inform personnel of the multiple hazards present in the lab, including the presence of radioactive material, cytotoxic compounds, and biological waste.
- Temperature range: ambient temperature
- Removable or permanent adhesive
- All labels are preprinted
Multi-purpose warning labels, ideal for offices, clinics, and laboratories. Paper warning labels feature a permanent adhesive to ensure they won't wrinkle or peel, and are designed for use at ambient temperatures. These general-use safety labels are fully customizable with logos, images, and alphanumeric text, in order to display appropriate warning information.
8 Options Available Below
Product Details & Documents
Various warning and safety labels for use throughout the lab. Designed to inform personnel of the multiple hazards present in the lab, including the presence of radioactive material, cytotoxic compounds, and biological waste.
- Temperature range: ambient temperature
- Removable or permanent adhesive
- All labels are preprinted
Multi-purpose warning labels, ideal for offices, clinics, and laboratories. Paper warning labels feature a permanent adhesive to ensure they won't wrinkle or peel, and are designed for use at ambient temperatures. These general-use safety labels are fully customizable with logos, images, and alphanumeric text, in order to display appropriate warning information.
Product Family Options
Informations sur le produit
- DésignationColorCore SizeDimensionsPackagingAvailabilityPrice
- Warning labels - CYTOTOXICRed/white on black3"25.4×38 mm (1×1.5")1000 labels/roll (1 label across)Log in pour la disponibilitéRoll of 1$59.59Supplier Number: 807656026664
- Warning labels - CYTOTOXICRed/white on black3"50.8×50.8 mm (2×2")1000 labels/roll (1 label across)Log in pour la disponibilitéRoll of 1$105.64Supplier Number: 807656026688
- Warning labels - CYTOTOXICRed/white on black3"101.6×101.6 mm (4×4")500 labels/roll (1 label across)Log in pour la disponibilitéRoll of 1$210.83Supplier Number: 807656026695
- Warning labels - RADIOACTIVERed on yellow3"69×25.4 mm (2.72×1")1000 labels/roll (1 label across)Log in pour la disponibilitéRoll of 1$78.58Supplier Number: 807656026718
- Warning labels - RADIOACTIVERed on yellow3"69×25.4 mm (2.72×1")1000 labels/roll (1 label across)Log in pour la disponibilitéRoll of 1$86.75Supplier Number: 807656026701
- Warning labels - R & D Use OnlyRed on yellow3"25.4×50.8 mm (1×2")500 labels/roll (1 label across)Log in pour la disponibilitéRoll of 1$78.58Supplier Number: 807656026749
- Warning labels - Calibrate Before UseRed on yellow3"25.4×50.8 mm (1×2")500 labels/roll (1 label across)Log in pour la disponibilitéRoll of 1$78.58Supplier Number: 807656026756
- Warning labels - Globally Harmonized SystemRed/white on black3"101.6×88.9 mm (4×3.5")500 labels/roll (1 label across)Log in pour la disponibilitéRoll of 1$113.67Supplier Number: 807656026770