Writable Cryogenic Multi-Color Labels for Microtubes Tops
Writable Cryogenic Multi-Color Labels for Microtubes Tops
Writable Cryogenic Multi-Color Labels for Microtubes Tops
Certificates
Writable Cryogenic Multi-Color Labels for Microtubes Tops
Writable Cryogenic Multi-Color Labels for Microtubes Tops
12 Options Available Below
Product Details & Documents
Writable cryo color dots and rectangles for identifying the sides and tops of cryo vials and microtubes stored in lab freezers and liquid nitrogen dewars. Easy to write on with permanent cryo markers. JTA-class labels are available in a variety of colors.
  • Temperature range: −196 to +110 °C (−321 to +230 °F)
  • Available in a range of colors for color-coding and easy identification
  • Writable rectangular labels for deep-freeze storage
  • Strong adhesive ensures labels won’t wrinkle or peel during long-term storage in freezers and on dry ice
Ideal for identification of vials, plastic bags, and other containers for long-term storage and color-coding in deep-freezing and harsh laboratory environments.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Dimensions
    Packaging
    Availability
    Price
  • Cryogenic writable labels for microtubes
    Blue
    26×10 mm (1×0.4")
    420 labels/pack (42 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $33.34
    $33.34
    Pack of 1
       $33.34
    Supplier Number: 807656067957
    Spécifications:
    More
    Specifications
    Cat. No.
    76275-704
  • Cryogenic writable labels for microtubes
    Beige
    26×10 mm (1×0.4")
    420 labels/pack (42 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $33.34
    $33.34
    Pack of 1
       $33.34
    Supplier Number: 807656067940
    Spécifications:
    More
    Specifications
    Cat. No.
    76275-690
  • Cryogenic writable labels for microtubes
    Bright green
    26×10 mm (1×0.4")
    420 labels/pack (42 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $33.34
    $33.34
    Pack of 1
       $33.34
    Supplier Number: 807656067971
    Spécifications:
    More
    Specifications
    Cat. No.
    76275-696
  • Cryogenic writable labels for microtubes
    Yellow
    26×10 mm (1×0.4")
    420 labels/pack (42 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $33.34
    $33.34
    Pack of 1
       $33.34
    Supplier Number: 807656068008
    Spécifications:
    More
    Specifications
    Cat. No.
    76275-694
  • Cryogenic writable labels for microtubes
    Mint
    26×10 mm (1×0.4")
    420 labels/pack (42 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $33.34
    $33.34
    Pack of 1
       $33.34
    Supplier Number: 807656068015
    Spécifications:
    More
    Specifications
    Cat. No.
    76275-698
  • Cryogenic writable labels for microtubes
    Dark brown
    26×10 mm (1×0.4")
    420 labels/pack (42 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $33.34
    $33.34
    Pack of 1
       $33.34
    Supplier Number: 807656067964
    Spécifications:
    More
    Specifications
    Cat. No.
    76275-708
  • Cryogenic writable labels for microtubes
    Orange
    26×10 mm (1×0.4")
    420 labels/pack (42 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $33.34
    $33.34
    Pack of 1
       $33.34
    Supplier Number: 807656068022
    Spécifications:
    More
    Specifications
    Cat. No.
    76275-706
  • Cryogenic writable labels for microtubes
    Red
    26×10 mm (1×0.4")
    420 labels/pack (42 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $33.34
    $33.34
    Pack of 1
       $33.34
    Supplier Number: 807656068046
    Spécifications:
    More
    Specifications
    Cat. No.
    76275-710
  • Cryogenic writable labels for microtubes
    Silver
    26×10 mm (1×0.4")
    420 labels/pack (42 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $33.34
    $33.34
    Pack of 1
       $33.34
    Supplier Number: 807656068053
    Spécifications:
    More
    Specifications
    Cat. No.
    76275-688
  • Cryogenic writable labels for microtubes
    Violet
    26×10 mm (1×0.4")
    420 labels/pack (42 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $33.34
    $33.34
    Pack of 1
       $33.34
    Supplier Number: 807656068060
    Spécifications:
    More
    Specifications
    Cat. No.
    76275-712
  • Cryogenic writable labels for microtubes
    White
    26×10 mm (1×0.4")
    420 labels/pack (42 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $33.34
    $33.34
    Pack of 1
       $33.34
    Supplier Number: 807656068077
    Spécifications:
    More
    Specifications
    Cat. No.
    76275-686
  • Cryogenic writable labels for microtubes
    Yellow
    26×10 mm (1×0.4")
    420 labels/pack (42 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $33.34
    $33.34
    Pack of 1
       $33.34
    Supplier Number: 807656068084
    Spécifications:
    More
    Specifications
    Cat. No.
    76275-692