Cryogenic Color Dots Writable Labels
Cryogenic Color Dots Writable Labels
Certificates
Cryogenic Color Dots Writable Labels
Cryogenic Color Dots Writable Labels
56 Options Available Below
Product Details & Documents
These small diameter, round cryo labels for microtube tops are made with a strong adhesive. Ideal for −80 °C freezers, dry ice, and liquid nitrogen storage.
  • Temperature range: −196 to +110 °C (−321 to +230 °F)
  • Available in a wide range of colors, for color-coding and easy identification
  • Easy to write on with permanent markers
  • Small round labels are ideal for 0.5 to 1.5 ml Eppendorf tube caps and vials
  • Strong adhesive ensures labels won't wrinkle or peel during long-term storage
  • Resistant to gamma irradiation (tested to up to 50 kGy)
Specially designed to comfortably fit 0.5, 0.6, and 1.5 ml Eppendorf tube caps and microtubes, these circular, waterproof cryo dot labels have a permanent adhesive that can withstand prolonged storage inside ultra-low temperature freezers. The freezer dot labels are supplied as Hagaki sheets and are available in 15 individual colors, as well as packs of 15 assorted colors for color-coding and rapid identification.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Dimensions
    Packaging
    Availability
    Price
  • Cryogenic color dots writable label
    Silver
    9 mm (0.354")
    520 labels/pack (104 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656024905
  • Cryogenic color dots writable label
    Beige
    9 mm (0.354")
    520 labels/pack (104 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656024912
  • Cryogenic color dots writable label
    Yellow
    9 mm (0.354")
    520 labels/pack (104 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656024929
  • Cryogenic color dots writable label
    Yellow
    9 mm (0.354")
    520 labels/pack (104 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656024936
  • Cryogenic color dots writable label
    Violet
    9 mm (0.354")
    520 labels/pack (104 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656024943
  • Cryogenic color dots writable label
    Mint green
    9 mm (0.354")
    520 labels/pack (104 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656000060
  • Cryogenic color dots writable label
    Bright green
    9 mm (0.354")
    520 labels/pack (104 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656024950
  • Cryogenic color dots writable label
    Pink
    9 mm (0.354")
    520 labels/pack (104 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $25.55
    $25.55
    Prix promotionnel pour les nouveaux clients
    Pack of 1
       $25.55
    Supplier Number: 807656024967
  • Cryogenic color dots writable label
    Lavender
    9 mm (0.354")
    520 labels/pack (104 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656024974
  • Cryogenic color dots writable label
    Blue
    9 mm (0.354")
    520 labels/pack (104 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $27.15
    $27.15
    Pack of 1
       $27.15
    Supplier Number: 807656024981
  • Cryogenic color dots writable label
    Orange
    9 mm (0.354")
    520 labels/pack (104 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $23.49
    $23.49
    Prix promotionnel pour les nouveaux clients
    Pack of 1
       $23.49
    Supplier Number: 807656024998
  • Cryogenic color dots writable label
    Dark brown
    9 mm (0.354")
    520 labels/pack (104 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656025001
  • Cryogenic color dots writable label
    White
    9 mm (0.354")
    520 labels/pack (104 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $27.15
    $27.15
    Pack of 1
       $27.15
    Supplier Number: 807656024899
  • Cryogenic color dots writable label
    Red
    9 mm (0.354")
    520 labels/pack (104 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $27.15
    $27.15
    Pack of 1
       $27.15
    Supplier Number: 807656025018
  • Cryogenic color dots writable label
    Light green
    9 mm (0.354")
    520 labels/pack (104 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656025025
  • Cryogenic color dots writable label, 15 colors
    Assorted
    9 mm (0.354")
    1560 labels/pack (104 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $81.56
    $81.56
    Pack of 1
       $81.56
    Supplier Number: 807656025032
  • Cryogenic color dots writable label, 8 colors
    Assorted
    9 mm (0.354")
    520 labels/pack (104 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $39.55
    $39.55
    Pack of 1
       $39.55
    Supplier Number: 807656003498
  • Cryogenic color dots writable label
    White
    11 mm (0.44")
    462 labels/pack (77 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $27.15
    $27.15
    Pack of 1
       $27.15
    Supplier Number: 807656025049
  • Cryogenic color dots writable label
    Silver
    11 mm (0.44")
    462 labels/pack (77 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656025056
  • Cryogenic color dots writable label
    Beige
    11 mm (0.44")
    462 labels/pack (77 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656025063
  • Cryogenic color dots writable label
    Yellow
    11 mm (0.44")
    462 labels/pack (77 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656025070
  • Cryogenic color dots writable label
    Yellow
    11 mm (0.44")
    462 labels/pack (77 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656025087
  • Cryogenic color dots writable label
    Bright green
    11 mm (0.44")
    462 labels/pack (77 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656025094
  • Cryogenic color dots writable label
    Mint green
    11 mm (0.44")
    462 labels/pack (77 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656025100
  • Cryogenic color dots writable label
    Pink
    11 mm (0.44")
    462 labels/pack (77 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656025117
  • Cryogenic color dots writable label
    Lavender
    11 mm (0.44")
    462 labels/pack (77 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656025124
  • Cryogenic color dots writable label
    Blue
    11 mm (0.44")
    462 labels/pack (77 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656025131
  • Cryogenic color dots writable label
    Orange
    11 mm (0.44")
    462 labels/pack (77 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656025148
  • Cryogenic color dots writable label
    Dark brown
    11 mm (0.44")
    462 labels/pack (77 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656025155
  • Cryogenic color dots writable label
    Red
    11 mm (0.44")
    462 labels/pack (77 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656025162
  • Cryogenic color dots writable label
    Violet
    11 mm (0.44")
    462 labels/pack (77 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656025179
  • Cryogenic color dots writable label
    Light green
    11 mm (0.44")
    462 labels/pack (77 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656025186
  • Cryogenic color dots writable label, 15 colors
    Assorted
    11 mm (0.44")
    1155 labels/pack (77 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $64.57
    $64.57
    Prix promotionnel pour les nouveaux clients
    Pack of 1
       $64.57
    Supplier Number: 807656025193
  • Cryogenic color dots writable label, 8 colors
    Assorted
    11 mm (0.44")
    462 labels/pack (77 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $39.55
    $39.55
    Pack of 1
       $39.55
    Supplier Number: 807656025209
  • Cryogenic color dots writable label
    White
    12.7 mm (0.5")
    420 labels/pack (60 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656000336
  • Cryogenic color dots writable label
    Silver
    12.7 mm (0.5")
    420 labels/pack (60 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656000473
  • Cryogenic color dots writable label
    Beige
    12.7 mm (0.5")
    420 labels/pack (60 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656000343
  • Cryogenic color dots writable label
    Yellow
    12.7 mm (0.5")
    420 labels/pack (60 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656000350
  • Cryogenic color dots writable label
    Yellow
    12.7 mm (0.5")
    420 labels/pack (60 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656000367
  • Cryogenic color dots writable label
    Bright green
    12.7 mm (0.5")
    420 labels/pack (60 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656000374
  • Cryogenic color dots writable label
    Mint green
    12.7 mm (0.5")
    420 labels/pack (60 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656000381
  • Cryogenic color dots writable label
    Pink
    12.7 mm (0.5")
    420 labels/pack (60 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.39
    $26.39
    Pack of 1
       $26.39
    Supplier Number: 807656000398
  • Cryogenic color dots writable label
    Lavender
    12.7 mm (0.5")
    420 labels/pack (60 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656000404
  • Cryogenic color dots writable label
    Blue
    12.7 mm (0.5")
    420 labels/pack (60 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656000411
  • Cryogenic color dots writable label
    Orange
    12.7 mm (0.5")
    420 labels/pack (60 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656000428
  • Cryogenic color dots writable label
    Dark brown
    12.7 mm (0.5")
    420 labels/pack (60 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656000435
  • Cryogenic color dots writable label
    Red
    12.7 mm (0.5")
    420 labels/pack (60 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656000442
  • Cryogenic color dots writable label
    Violet
    12.7 mm (0.5")
    420 labels/pack (60 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656000459
  • Cryogenic color dots writable label
    Light green
    12.7 mm (0.5")
    420 labels/pack (60 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $26.89
    $26.89
    Pack of 1
       $26.89
    Supplier Number: 807656000466
  • Cryogenic color dots writable label, 15 colors
    Assorted
    12.7 mm (0.5")
    900 labels/pack (60 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $58.27
    $58.27
    Pack of 1
       $58.27
    Supplier Number: 807656000480
  • Cryogenic color dots writable label
    White
    19 mm (0.75")
    420 labels/pack (21 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $38.08
    $38.08
    Pack of 1
       $38.08
    Supplier Number: 807656025230
  • Cryogenic color dots writable label
    Pink
    19 mm (0.75")
    420 labels/pack (21 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $38.08
    $38.08
    Pack of 1
       $38.08
    Supplier Number: 807656025247
  • Cryogenic color dots writable label
    Red
    19 mm (0.75")
    420 labels/pack (21 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $38.08
    $38.08
    Pack of 1
       $38.08
    Supplier Number: 807656025254
  • Cryogenic color dots writable label
    Yellow
    19 mm (0.75")
    420 labels/pack (21 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $38.08
    $38.08
    Pack of 1
       $38.08
    Supplier Number: 807656025261
  • Cryogenic color dots writable label
    Blue
    19 mm (0.75")
    420 labels/pack (21 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $38.08
    $38.08
    Pack of 1
       $38.08
    Supplier Number: 807656025278
  • Cryogenic color dots writable label
    Light green
    19 mm (0.75")
    420 labels/pack (21 labels/sheet)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $38.08
    $38.08
    Pack of 1
       $38.08
    Supplier Number: 807656025285