TitroLine® 5000 Titrators, SI Analytics
 :   YSI
TitroLine® 5000 Titrators, SI Analytics
 :  YSI
TitroLine® 5000 Titrators, SI Analytics
The TitroLine® 5000, successor of the TitroLine® easy, is a very easy to use automatic titrator for a variety of applications, including water/wastewater applications, environmental and food analysis.
  • High resolution pH/mV measurement input for pH, ORP, silver and further mV electrodes
  • PT1000 and NTC 30 temperature measurement input for automatic temperature compensation
  • Pre-installed standard methods for FOS/TAC, alkalinity, total acidity in drinks, chloride
  • Linear and dynamic titration to equivalence point
  • Titration to pH and mV endpoints
  • Same manual titration and dosing function as the Titronic® 300 piston burette
 : Basic (M1) units supplied with pre-assembled 20 or 50 ml dosing unit, manual controller TZ 3880, titration clamp, stand rod, magnetic stirrer TM 50 and power supply. M2 units also include a pH electrode and buffer set. The M3 unit includes an Ag electrode instead of a pH electrode.
  • Catalog No:
  • 76403-684
  • 76403-686
  • 76403-694
  • 76403-696
  • Model:
  • TitroLine® 5000
  • TitroLine® 5000
  • TitroLine® 5000
  • TitroLine® 5000
  • Display:
  • Graphical TFT
  • Graphical TFT
  • Graphical TFT
  • Graphical TFT
  • Display resolution:
  • 0.005 - 0.025 ml (depending on dosing unit)
  • 0.005 - 0.025 ml (depending on dosing unit)
  • 0.005 - 0.025 ml (depending on dosing unit)
  • 0.005 - 0.025 ml (depending on dosing unit)
  • Volume display:
  • 0000.000 - 9999.999 ml
  • 0000.000 - 9999.999 ml
  • 0000.000 - 9999.999 ml
  • 0000.000 - 9999.999 ml
  • Keypad:
  • Optional USB keyboard (TZ 3835) can be connected
  • Optional USB keyboard (TZ 3835) can be connected
  • Optional USB keyboard (TZ 3835) can be connected
  • Optional USB keyboard (TZ 3835) can be connected
  • Stirrer connection:
  • Connection for stirrer TM 50 (included)
  • Connection for stirrer TM 50 (included)
  • Connection for stirrer TM 50 (included)
  • Connection for stirrer TM 50 (included)
  • Dosing accuracy:
  • 0.15% (systematic error); 0.05% (random error)
  • 0.15% (systematic error); 0.05% (random error)
  • 0.15% (systematic error); 0.05% (random error)
  • 0.15% (systematic error); 0.05% (random error)
  • Dosing volume:
  • 20/50 ml
  • 20/50 ml
  • 20/50 ml
  • 20/50 ml
  • Dosing speed:
  • Max. 100 ml/min (with 50 ml unit)
  • Max. 100 ml/min (with 50 ml unit)
  • Max. 100 ml/min (with 50 ml unit)
  • Max. 100 ml/min (with 50 ml unit)
  • Filling time:
  • 30 s - 999 s (time according to the cylinder volume)
  • 30 s - 999 s (time according to the cylinder volume)
  • 30 s - 999 s (time according to the cylinder volume)
  • 30 s - 999 s (time according to the cylinder volume)
  • Burette resolution:
  • 8000 steps
  • 8000 steps
  • 8000 steps
  • 8000 steps
  • Measuring input electrode:
  • pH/mV input with electrode socket according to DIN 19 262
  • pH/mV input with electrode socket according to DIN 19 262
  • pH/mV input with electrode socket according to DIN 19 262
  • pH/mV input with electrode socket according to DIN 19 262
  • mV accuracy:
  • −1900 to 1900…1.0 ±1
  • −1900 to 1900…1.0 ±1
  • −1900 to 1900…1.0 ±1
  • −1900 to 1900…1.0 ±1
  • Measuring input PT1000:
  • Socket 2×4 mm (PT1000/NTC 30)
  • Socket 2×4 mm (PT1000/NTC 30)
  • Socket 2×4 mm (PT1000/NTC 30)
  • Socket 2×4 mm (PT1000/NTC 30)
  • Connections:
  • 1× USB-A, 1× USB-B, 2× RS 232C
  • 1× USB-A, 1× USB-B, 2× RS 232C
  • 1× USB-A, 1× USB-B, 2× RS 232C
  • 1× USB-A, 1× USB-B, 2× RS 232C
  • Power supply:
  • 100 - 240 V, 50/60 Hz
  • 100 - 240 V, 50/60 Hz
  • 100 - 240 V, 50/60 Hz
  • 100 - 240 V, 50/60 Hz
  • Operating temperature:
  • 10...40 °C (50...104 °F)
  • 10...40 °C (50...104 °F)
  • 10...40 °C (50...104 °F)
  • 10...40 °C (50...104 °F)
  • Compliance:
  • ISO 8655, part 6; EMC: 2004/108/EG; safety EG Directive 2006/95
  • ISO 8655, part 6; EMC: 2004/108/EG; safety EG Directive 2006/95
  • ISO 8655, part 6; EMC: 2004/108/EG; safety EG Directive 2006/95
  • ISO 8655, part 6; EMC: 2004/108/EG; safety EG Directive 2006/95
  • W×D×H:
  • 13.5×20.5×31.0 cm
  • 13.5×20.5×31.0 cm
  • 13.5×20.5×31.0 cm
  • 13.5×20.5×31.0 cm
  • Weight:
  • 2 kg (4.4 lbs.)
  • 2 kg (4.4 lbs.)
  • 2 kg (4.4 lbs.)
  • 2 kg (4.4 lbs.)
Product Family Options
  • Désignation
  • TitroLine® 5000 M1 with 20 ml dosing unit and magnetic stirrer (TM 235)
    Each
    Each - $12,684.83
    $12,684.83
    Each
       $12,684.83
     : 285230100
    Specifications
    Cat. No.
    76403-684
    Model
    TitroLine® 5000
    Display
    Graphical TFT
    Display resolution
    0.005 - 0.025 ml (depending on dosing unit)
    Volume display
    0000.000 - 9999.999 ml
    Keypad
    Optional USB keyboard (TZ 3835) can be connected
    Stirrer connection
    Connection for stirrer TM 50 (included)
    Dosing accuracy
    0.15% (systematic error); 0.05% (random error)
    Dosing volume
    20/50 ml
    Dosing speed
    Max. 100 ml/min (with 50 ml unit)
    Filling time
    30 s - 999 s (time according to the cylinder volume)
    Burette resolution
    8000 steps
    Measuring input electrode
    pH/mV input with electrode socket according to DIN 19 262
    mV accuracy
    −1900 to 1900…1.0 ±1
    Measuring input PT1000
    Socket 2×4 mm (PT1000/NTC 30)
    Connections
    1× USB-A, 1× USB-B, 2× RS 232C
    Power supply
    100 - 240 V, 50/60 Hz
    Operating temperature
    10...40 °C (50...104 °F)
    Compliance
    ISO 8655, part 6; EMC: 2004/108/EG; safety EG Directive 2006/95
    W×D×H
    13.5×20.5×31.0 cm
    Weight
    2 kg (4.4 lbs.)
  • TitroLine® 5000 M1 with 50 ml dosing unit and magnetic stirrer (TM 235)
    Each
    Each - $11,882.33
    $11,882.33
    Each
       $11,882.33
     : 285230150
    Specifications
    Cat. No.
    76403-686
    Model
    TitroLine® 5000
    Display
    Graphical TFT
    Display resolution
    0.005 - 0.025 ml (depending on dosing unit)
    Volume display
    0000.000 - 9999.999 ml
    Keypad
    Optional USB keyboard (TZ 3835) can be connected
    Stirrer connection
    Connection for stirrer TM 50 (included)
    Dosing accuracy
    0.15% (systematic error); 0.05% (random error)
    Dosing volume
    20/50 ml
    Dosing speed
    Max. 100 ml/min (with 50 ml unit)
    Filling time
    30 s - 999 s (time according to the cylinder volume)
    Burette resolution
    8000 steps
    Measuring input electrode
    pH/mV input with electrode socket according to DIN 19 262
    mV accuracy
    −1900 to 1900…1.0 ±1
    Measuring input PT1000
    Socket 2×4 mm (PT1000/NTC 30)
    Connections
    1× USB-A, 1× USB-B, 2× RS 232C
    Power supply
    100 - 240 V, 50/60 Hz
    Operating temperature
    10...40 °C (50...104 °F)
    Compliance
    ISO 8655, part 6; EMC: 2004/108/EG; safety EG Directive 2006/95
    W×D×H
    13.5×20.5×31.0 cm
    Weight
    2 kg (4.4 lbs.)
  • TitroLine® 5000 with 20 ml dosing unit and magnetic stirrer, certification included
    Each
    Each - $12,570.17
    $12,570.17
    Each
       $12,570.17
     : 285230610
    Specifications
    Cat. No.
    76403-694
    Model
    TitroLine® 5000
    Display
    Graphical TFT
    Display resolution
    0.005 - 0.025 ml (depending on dosing unit)
    Volume display
    0000.000 - 9999.999 ml
    Keypad
    Optional USB keyboard (TZ 3835) can be connected
    Stirrer connection
    Connection for stirrer TM 50 (included)
    Dosing accuracy
    0.15% (systematic error); 0.05% (random error)
    Dosing volume
    20/50 ml
    Dosing speed
    Max. 100 ml/min (with 50 ml unit)
    Filling time
    30 s - 999 s (time according to the cylinder volume)
    Burette resolution
    8000 steps
    Measuring input electrode
    pH/mV input with electrode socket according to DIN 19 262
    mV accuracy
    −1900 to 1900…1.0 ±1
    Measuring input PT1000
    Socket 2×4 mm (PT1000/NTC 30)
    Connections
    1× USB-A, 1× USB-B, 2× RS 232C
    Power supply
    100 - 240 V, 50/60 Hz
    Operating temperature
    10...40 °C (50...104 °F)
    Compliance
    ISO 8655, part 6; EMC: 2004/108/EG; safety EG Directive 2006/95
    W×D×H
    13.5×20.5×31.0 cm
    Weight
    2 kg (4.4 lbs.)
  • TitroLine® 5000 with 50 ml dosing unit and magnetic stirrer, certification included
    Each
    Each - $12,570.17
    $12,570.17
    Each
       $12,570.17
     : 285230620
    Specifications
    Cat. No.
    76403-696
    Model
    TitroLine® 5000
    Display
    Graphical TFT
    Display resolution
    0.005 - 0.025 ml (depending on dosing unit)
    Volume display
    0000.000 - 9999.999 ml
    Keypad
    Optional USB keyboard (TZ 3835) can be connected
    Stirrer connection
    Connection for stirrer TM 50 (included)
    Dosing accuracy
    0.15% (systematic error); 0.05% (random error)
    Dosing volume
    20/50 ml
    Dosing speed
    Max. 100 ml/min (with 50 ml unit)
    Filling time
    30 s - 999 s (time according to the cylinder volume)
    Burette resolution
    8000 steps
    Measuring input electrode
    pH/mV input with electrode socket according to DIN 19 262
    mV accuracy
    −1900 to 1900…1.0 ±1
    Measuring input PT1000
    Socket 2×4 mm (PT1000/NTC 30)
    Connections
    1× USB-A, 1× USB-B, 2× RS 232C
    Power supply
    100 - 240 V, 50/60 Hz
    Operating temperature
    10...40 °C (50...104 °F)
    Compliance
    ISO 8655, part 6; EMC: 2004/108/EG; safety EG Directive 2006/95
    W×D×H
    13.5×20.5×31.0 cm
    Weight
    2 kg (4.4 lbs.)