DNP Buffer (Neutralizing Fluid), Hardy Diagnostics
DNP Buffer, 200 ml, Glass bottle, Boston round
Supplier #: U377
Recommended for neutralizing the activity of antimicrobial agents according to the European Pharmacopoeia.
- Type de conditionnement:Glass bottle, Boston round
- Désignation:DNP Buffer
- Taille:200 ml