Removable deep freeze labels for vials, tubes, and freezer boxes. These thermal transfer freezer labels have a glove-friendly adhesive that ensures the labels can be easily removed without leaving any adhesive residue.
- Temperature range: –80 to 100 ℃ (–112 to 212 ℉)
- Ideal for long-term storage in deep-freeze conditions (–80 ℃)
- Removable adhesive ensures the labels can be easily removed without leaving any adhesive residue
- Printable with all major thermal-transfer printer brands
- Resistant to gamma irradiation (tested up to 50 KGy)
Deep freeze, removable barcode labels for vials, tubes, as well as polypropylene and cardboard boxes. The freezer labels have a strong, glove-friendly adhesive that allows them to adhere firmly to containers in (–20 ℃, –40 ℃, –80 ℃) freezers and dry ice while still being easily removable at room temperature without leaving behind any residue.
Waterproof and tearproof, these removable deep freeze labels can also withstand temperatures ranging up to 100 ℃, making them suitable for general lab use such as the identification of beakers, flasks, boxes, plastic and glass bottles that are stored in fridges and freezers.
Removable deep freeze labels for vials, tubes, and freezer boxes. These thermal transfer freezer labels have a glove-friendly adhesive that ensures the labels can be easily removed without leaving any adhesive residue.
- Temperature range: –80 to 100 ℃ (–112 to 212 ℉)
- Ideal for long-term storage in deep-freeze conditions (–80 ℃)
- Removable adhesive ensures the labels can be easily removed without leaving any adhesive residue
- Printable with all major thermal-transfer printer brands
- Resistant to gamma irradiation (tested up to 50 KGy)
Deep freeze, removable barcode labels for vials, tubes, as well as polypropylene and cardboard boxes. The freezer labels have a strong, glove-friendly adhesive that allows them to adhere firmly to containers in (–20 ℃, –40 ℃, –80 ℃) freezers and dry ice while still being easily removable at room temperature without leaving behind any residue.
Waterproof and tearproof, these removable deep freeze labels can also withstand temperatures ranging up to 100 ℃, making them suitable for general lab use such as the identification of beakers, flasks, boxes, plastic and glass bottles that are stored in fridges and freezers.
Product Family Options
Circle removable deep freeze labels for thermal transfer printers
- DésignationColorCore SizeDimensionsPackaging
- Deep freeze labelWhite1"19.1 mm (0.75")2000 labels/roll (1 label across)Roll of 1$221.88: 807656016511
- Deep freeze labelWhite3"19.1 mm (0.75")2000 labels/roll (1 label across)Roll of 1$221.88: 807656016528
- Deep freeze labelWhite1"25.4 mm (1")1000 labels/roll (1 label across)Roll of 1$99.36: 807656016535
- Deep freeze labelWhite3"25.4 mm (1")5000 labels/roll (1 label across)Roll of 1$496.09: 807656016559
Rectangle and circle removable deep freeze labels for thermal transfer printers
- DésignationColorCore SizeDimensionsPackaging
- Deep freeze labelWhite1"23.9×12.7+11.1 mm (0.94×0.5+0.44")2000 labels/roll (set of 1 rectangle and 1 circle)Roll of 1$236.20: 807656016450
- Deep freeze labelWhite3"23.9×12.7+11.1 mm (0.94×0.5+0.44")4000 labels/roll (set of 1 rectangle and 1 circle)Roll of 1$472.66: 807656016467
- Deep freeze labelWhite1"28.6×19.1+11.1 mm (1.12×0.75+0.44")2000 labels/roll (set of 1 rectangle and 1 circle)Roll of 1$243.30: 807656016474
- Deep freeze labelWhite3"28.6×19.1+11.1 mm (1.12×0.75+0.44")4000 labels/roll (set of 1 rectangle and 1 circle)Roll of 1$486.85: 807656016481
- Deep freeze labelWhite1"30×7.1+9.4 mm (1.18×0.28+0.37")1000 labels/roll (set of 2 rectangles and 1 circle)Roll of 1$118.39: 807656016498
- Deep freeze labelWhite3"30×7.1+9.4 mm (1.18×0.28+0.37")1000 labels/roll (set of 2 rectangles and 1 circle)Roll of 1$354.44: 807656016504
Rectangle removable deep freeze labels for thermal transfer printers
- DésignationColorCore SizeDimensionsPackaging
- Deep freeze labelWhite1"6.4×25.4 mm (0.25×1")10000 labels/roll (10 labels across)Roll of 1$478.60: 807656015958
- Deep freeze labelWhite3"6.4×25.4 mm (0.25×1")10000 labels/roll (10 labels across)Roll of 1$478.60: 807656015965
- Deep freeze labelWhite1"22.2×11 mm (0.87×0.44")2000 labels/roll (1 label across)Roll of 1$160.50: 807656015972
- Deep freeze labelWhite3"22.2×11 mm (0.87×0.44")5000 labels/roll (1 label across)Roll of 1$397.14: 807656027623
- Deep freeze labelWhite1"22.2×203.2 mm (0.87×8")250 labels/roll (1 label across)Roll of 1$138.11: 807656015989
- Deep freeze labelWhite1"25.4×7 mm (1×0.28")2000 labels/roll (1 label across)Roll of 1$155.97: 807656015996
- Deep freeze labelWhite3"25.4×7 mm (1×0.28")4000 labels/roll (1 label across)Roll of 1$311.87: 807656016009
- Deep freeze labelWhite1"25.4×12.7 mm (1×0.5")1000 labels/roll (1 label across)Roll of 1$85.94: 807656016016
- Deep freeze labelWhite3"25.4×12.7 mm (1×0.5")3000 labels/roll (1 label across)Roll of 1$257.16: 807656016023
- Deep freeze labelWhite1"25.4×17.5 mm (1×0.69")1000 labels/roll (1 label across)Roll of 1$89.60: 807656016030
- Deep freeze labelWhite3"25.4×17.5 mm (1×0.69")1000 labels/roll (1 label across)Roll of 1$89.60: 807656016047
- Deep freeze labelWhite1"25.4×19.1 mm (1×0.75")1000 labels/roll (1 label across)Roll of 1$93.98: 807656016054
- Deep freeze labelWhite3"25.4×19.1 mm (1×0.75")5000 labels/roll (1 label across)Roll of 1$470.54: 807656016061
- Deep freeze labelWhite1"25.4×25.4 mm (1×1")2000 labels/roll (1 label across)Roll of 1$204.41: 807656016078
- Deep freeze labelWhite3"25.4×25.4 mm (1×1")2000 labels/roll (1 label across)Roll of 1$204.41: 807656016085
- Deep freeze labelWhite1"25.4×44.5 mm (1×1.75")1000 labels/roll (1 label across)Roll of 1$147.15: 807656016092
- Deep freeze labelWhite3"25.4×44.5 mm (1×1.75")3000 labels/roll (1 label across)Roll of 1$441.02: 807656016108
- Deep freeze labelWhite1"25.4×44.5 mm (1×1.75")3000 labels/roll (3 labels across)Roll of 1$441.21: 807656016115
- Deep freeze labelWhite3"25.4×44.5 mm (1×1.75")10000 labels/roll (3 labels across)Roll of 1$1,472.40: 807656016122
- Deep freeze labelWhite1"25.4×50.8 mm (1×2")1000 labels/roll (1 label across)Roll of 1$204.41: 807656016139
- Deep freeze labelWhite3"25.4×50.8 mm (1×2")1000 labels/roll (1 label across)Roll of 1$204.41: 807656016146
- Deep freeze labelWhite1"32.5×15.9 mm (1.28×0.62")1000 labels/roll (1 label across)Roll of 1$95.90: 807656016153
- Deep freeze labelWhite3"32.5×15.9 mm (1.28×0.62")1000 labels/roll (1 label across)Roll of 1$95.90: 807656016160
- Deep freeze labelWhite1"33×15.9 mm (1.3×0.62")1000 labels/roll (1 label across)Roll of 1$95.90: 807656016177
- Deep freeze labelWhite3"33×15.9 mm (1.3×0.62")1000 labels/roll (1 label across)Roll of 1$95.90: 807656016184
- Deep freeze labelWhite1"38.1×19.1 mm (1.5×0.75")1000 labels/roll (1 label across)Roll of 1,000$116.09: 807656031132
- Deep freeze labelWhite3"38.1×19.1 mm (1.5×0.75")2000 labels/roll (1 label across)Roll of 2,000$230.11: 807656031149
- Deep freeze labelWhite1"38.1×63.5 mm (1.5×2.5")1000 labels/roll (1 label across)Roll of 1$321.22: 807656016191
- Deep freeze labelWhite3"38.1×63.5 mm (1.5×2.5")1000 labels/roll (1 label across)Roll of 1$321.22: 807656016207
- Deep freeze labelWhite1"44.5×12.7 mm (1.75×0.5")2000 labels/roll (1 label across)Roll of 1$171.53: 807656016214
- Deep freeze labelWhite3"44.5×12.7 mm (1.75×0.5")6000 labels/roll (1 label across)Roll of 1$509.71: 807656016221
- Deep freeze labelWhite1"44.5×25.4 mm (1.75×1")2000 labels/roll (1 label across)Roll of 1$265.56: 807656016238
- Deep freeze labelWhite3"44.5×25.4 mm (1.75×1")2000 labels/roll (1 label across)Roll of 1$265.56: 807656016245
- Deep freeze labelWhite1"45.7×38.1 mm (1.8×1.5")250 labels/roll (1 label across)Roll of 1$152.86: 807656016412
- Deep freeze labelWhite3"45.7×38.1 mm (1.8×1.5")500 labels/roll (1 label across)Roll of 1$305.48: 807656016429
- Deep freeze labelWhite1"50.8×40.6 mm (2×1.6")1000 labels/roll (1 label across)Roll of 1$257.25: 807656016276
- Deep freeze labelWhite3"50.8×40.6 mm (2×1.6")1000 labels/roll (1 label across)Roll of 1$257.25: 807656016283
- Deep freeze labelWhite1"50.8×25.4 mm (2×1")1000 labels/roll (1 label across)Roll of 1$152.41: 807656001562
- Deep freeze labelOrange1"50.8×25.4 mm (2×1")1000 labels/roll (1 label across)Roll of 1$152.41: 807656118154
- Deep freeze labelOrange3"50.8×25.4 mm (2×1")1000 labels/roll (1 label across)Roll of 1$152.41: 807656118161
- Deep freeze labelWhite1"63.5×25.4 mm (2.5×1")1000 labels/roll (1 label across)Roll of 1$190.35: 807656016290
- Deep freeze labelYellow1"63.5×25.4 mm (2.5×1")1000 labels/roll (1 label across)Roll of 1$190.35: 807656016306
- Deep freeze labelBright green1"63.5×25.4 mm (2.5×1")1000 labels/roll (1 label across)Roll of 1$190.35: 807656016313
- Deep freeze labelRed1"63.5×25.4 mm (2.5×1")1000 labels/roll (1 label across)Roll of 1$190.35: 807656016320
- Deep freeze labelWhite3"63.5×25.4 mm (2.5×1")3000 labels/roll (1 label across)Roll of 1$570.96: 807656016337
- Deep freeze labelYellow3"63.5×25.4 mm (2.5×1")3000 labels/roll (1 label across)Roll of 1$570.96: 807656016344
- Deep freeze labelBright green3"63.5×25.4 mm (2.5×1")3000 labels/roll (1 label across)Roll of 1$570.96: 807656016351
- Deep freeze labelRed3"63.5×25.4 mm (2.5×1")3000 labels/roll (1 label across)Roll of 1$570.96: 807656016368
- Deep freeze labelWhite1"67.1×7 mm (2.64×0.28")2000 labels/roll (1 label across)Roll of 1$170.48: 807656016375
- Deep freeze labelWhite3"67.1×7 mm (2.64×0.28")4000 labels/roll (1 label across)Roll of 1$340.79: 807656016382
- Deep freeze labelWhite1"67.1×7 mm (2.64×0.28")4000 labels/roll (1 label across)Roll of 1$330.86: 807656016399
- Deep freeze labelWhite3"67.1×7 mm (2.64×0.28")4000 labels/roll (1 label across)Roll of 1$330.86: 807656016405
- Deep freeze labelWhite1"76.2×25.4 mm (3×1")1000 labels/roll (1 label across)Roll of 1$193.05: 807656001548
- Deep freeze labelWhite3"76.2×25.4 mm (3×1")1000 labels/roll (1 label across)Roll of 1$193.05: 807656001555
- Deep freeze labelWhite1"101.6×101.6 mm (4×4")300 labels/roll (1 label across)Roll of 1$271.50: 807656016436
- Deep freeze labelWhite3"101.6×101.6 mm (4×4")500 labels/roll (1 label across)Roll of 1$452.71: 807656016443