Thermo Sequenase DNA Polymerase, Cytiva
Supplier: Cytiva
Certificates
About this item
Thermo Sequenase is a novel thermostable DNA polymerase that uses dideoxynucleotide triphosphates (ddNTPs) as readily as deoxynucleotide (dNTP) substrates. This results in extremely uniform and easy-to-read sequence band patterns. It is an excellent choice for cycle sequencing where template quantity limitations preclude the use of T7 Sequenase DNA polymerase.
- One unit of enzyme incorporates 10 nmol of dNTPs into acid-insoluble material in 30 min at 74°C
- For DNA Sequencing and SNP genotyping applications where primer extension is employed
- Uniform and easy-to-read band patterns
- Thermostable enzyme
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together