Safety Vest, V2CL2ML Series, ANSI/ISEA Class 2, MCR Safety
This ANSI/ISEA Class 2 value high visibliity safety vest is made of fluorescent lime polyester mesh material.
- 2" silver reflective stripes
- Hook and loop front closure
- Cool mesh material, lime
- No pockets
Mesh construction offers increased breathability in warm environments.
: ANSI/ISEA 107 Type R, Class 2 compliant.
- Catalog No:
- 76339-952
- 76339-954
- 76340-130
- 76340-062
- 76340-028
- 76339-984
- Model:
- V2CL2ML
- V2CL2ML
- V2CL2ML
- V2CL2ML
- V2CL2ML
- V2CL2ML
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Gender:
- For Men
- For Men
- For Men
- For Men
- For Men
- For Men
- Sleeve or leg length:
- Short
- Short
- Short
- Short
- Short
- Short
- Closure type:
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Fabric:
- Polyester mesh
- Polyester mesh
- Polyester mesh
- Polyester mesh
- Polyester mesh
- Polyester mesh
Frequently Bought Together