Paper Labels, Blank, 3" Circle, Labelmaster
Supplier: Label Master
Certificates
About this item
The labeling possibilities are virtually endless with Blank 3" Circle Paper labels.
- Thermal transfer imprintable
- Outstanding chemical and abrasion resistance
- Strong adhesion to corrugated boxes
Blank labels may be used for countless applications, from identifying inventory and property to marking packages with special handling information.
Frequently Bought Together