KIMBLE® MICROFLEX® Kjeldal-Shaped Flasks, Threaded, DWK Life Sciences
KIMBLE® MICROFLEX® Kjeldal-Shaped Flasks, Threaded, DWK Life Sciences
KIMBLE® MICROFLEX® Kjeldal-Shaped Flasks, Threaded, DWK Life Sciences
Certificates
KIMBLE® MICROFLEX® Kjeldal-Shaped Flasks, Threaded, DWK Life Sciences
KIMBLE® MICROFLEX® Kjeldal-Shaped Flasks, Threaded, DWK Life Sciences
2 Options Available Below
Product Details & Documents
This flask has a conventional round bottom shape with a tapered upper portion for easy recovery of reaction products.
  • Accessory for Microscale MICROFLEX® Standard Taper 14/10 threaded kits
Manufactured from 33 expansion, low extractable borosilicate glass.
 : Glass conforms to USP Type I and ASTM E438, Type I, Class A requirements.
Product Family Options
Product Information
  • Capacité
    Désignation
    Availability
    Price
  • 10 ml
    Recovery flask, 20-400
    Log in for availability
    Log in for availability
    Each
    Each - $51.60
    $51.60
    New Customer Promotional Price
    Each
       $51.60
    Supplier Number: 747550-1020
  • 25 ml
    Recovery flask, 20-400
    Log in for availability
    Log in for availability
    Each
    Each - $65.70
    $65.70
    New Customer Promotional Price
    Each
       $65.70
    Supplier Number: 747550-2520