Maintenance Solutions for Electrodes
Maintenance Solutions for Electrodes
Supplier:  Mettler Toledo
Certificates
Maintenance Solutions for Electrodes
Maintenance Solutions for Electrodes
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This solution keeps your electrode ready with the right cleaning, storage and regeneration solution.
 : Includes quality inspection certificate.
 : Packaging type - bottle.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • Cleaning solution thiourea, colorles, odorless, completely miscible in water
    250 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $53.80
    $53.80
    Each
       $53.80
    Supplier Number: 51350102
  • InLab storage solution
    250 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $64.30
    $64.30
    Each(1/Itm/Bx)
       $64.30
    Supplier Number: 30111142