WHEATON® Replacement Caps for Valumetric™ Graduated Bottles, DWK Life Sciences
10847-612
- Cap Size:38-400
- Cat. no.:10847-612
- Supplier No.:W240541
High-quality replacement caps for Valumetric™ graduated bottles.
- Stress crack resistant
- Superior torque retention
- Excellent sealing characteristics
Manufactured from LDPE. The unique cone design provides a wedge-type seal that not only seals across the top, but also across the inside diameter.