Stainless Steel Lab Scoops, Eisco Scientific
470219-130
: Wards
- Description:Stainless steel lab scoops
- Cat. no.:470219-130
- Supplier No.:CH0635A
Durable and versatile lab scoops for classroom or laboratory.
- Lab scoops/spatulas
- 6.5" long
Many uses in science laboratories and classrooms. These stainless steel scoops features one pointed end, one rounded end, and semicircular cross section.