ContactUsLinkComponent
EMS Kuehne Coverglass/Specimen Forceps, Electron Microscopy Sciences
Catalog # 76375-964
undefined
EMS Kuehne Coverglass/Specimen Forceps, Electron Microscopy Sciences
Catalog # 76375-964
Supplier Number:  72997-07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Kuehne coverglass/specimen forceps
  • Length:
    107 mm
  • Cat. no.:
    76375-964
Specifications
About this item
Forceps specifically designed for handling microscope slides.
  • Made of stainless steel