Autosampler Vials, Headspace, DWK Life Sciences | Avantor
Autosampler Vials, Headspace, DWK Life Sciences
Autosampler Vials, Headspace, DWK Life Sciences
Certificates
Autosampler Vials, Headspace, DWK Life Sciences
Autosampler Vials, Headspace, DWK Life Sciences
2 Options Available Below
Product Details & Documents
Round bottom vials distribute the internal pressure created at high temperatures across the glass surface and are more easily handled by robotic arms that lift the vial from the tray.
  • Designed to fit most headspace autosamplers
  • Flat finish on top and bottom to maximise heating efficiency
The beveled edge 20 mm crimp seal finish features a sturdy rim that presses into the septum for a more effective seal.
Certifications: Meets or exceeds OEM specifications.
Product Family Options
Informations sur le produit
  • Désignation
    Diameter ×H
    Color
    Avec bouchon
    Availability
    Price
  • Sample vial with modified top
    23×46 mm
    Clear
    Non
    Discontinued
    Supplier Number: 90S332346B
  • Sample vial with modified top
    23×75 mm
    Clear
    Non
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $802.62
    $802.62
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $802.62
    Supplier Number: 90S332375BMB
  • Sample vial with modified top and bottom
    23×75 mm
    Clear
    Non
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $802.62
    $802.62
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $802.62
    Supplier Number: 90S332375BMB