Ergonomic Paddle Handle Forceps, Electron Microscopy Sciences
Marketsource
Ergonomic Paddle Handle Forceps, Electron Microscopy Sciences
Marketsource
Certificates
Ergonomic Paddle Handle Forceps, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Conventional forceps made of premium grade, hardened stainless steel with a protective coating for extra durability and protection from rust and corrosion.
  • Satin-matte finish reduces eye-fatiguing glare
  • Available in a variety of length, tip, and handle styles
Curved styles are well suited for retreiving specimens from the inside perimeter of transport jars and tissue separation in the water bath.
Product Family Options
Paddle Handle Forceps
Informations sur le produit
  • Désignation
    Overall Length
    Tip Length
    Tip Configuration
    Availability
    Price
  • Ergonomic forceps, paddle handle
    178 mm
    3 mm
    Smooth tips
    Discontinued
    Supplier Number: 77937-44
    Spécifications:
    Marketsource
    Marketsource
  • Ergonomic forceps, paddle handle
    178 mm
    3 mm
    Serrated tips
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $373.69
    $373.69
    Each
       $373.69
    Supplier Number: 77937-45
  • Ergonomic forceps, paddle handle
    178 mm
    3 mm
    Tips with 1×2 teeth
    Discontinued
    Supplier Number: 77937-46
    Spécifications:
    Marketsource
    Marketsource
  • Ergonomic forceps, paddle handle
    178 mm
    1.5 mm
    Smooth tips
    Discontinued
    Supplier Number: 77937-47
    Spécifications:
    Marketsource
    Marketsource
  • Ergonomic forceps, paddle handle
    178 mm
    1.5 mm
    Serrated tips
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $356.44
    $356.44
    Each
       $356.44
    Supplier Number: 77937-48
  • Ergonomic forceps, paddle handle
    178 mm
    Needle tip
    Discontinued
    Supplier Number: 77937-49
    Spécifications:
    Marketsource
    Marketsource
  • Ergonomic forceps, paddle handle
    178 mm
    1.5 mm
    Curved serrated tips
    Discontinued
    Supplier Number: 77937-54
    Spécifications:
    Marketsource
    Marketsource
  • Ergonomic forceps, paddle handle
    178 mm
    1.5 mm
    Curved smooth tips
    Discontinued
    Supplier Number: 77937-55
    Spécifications:
    Marketsource
    Marketsource