ContactUsLinkComponent
KIMBLE® AOCS Color Comparison Tube, DWK Life Sciences
Catalog # 66176-005
KIMBLE® AOCS Color Comparison Tube, DWK Life Sciences
Catalog # 66176-005
Supplier Number:  45290-154
Specifications
  • Description:
    AOCS color comparison tubes, I.D. 19 mm, distance rings from inside bottom 25.4, 67, 133 mm
  • Cat. no.:
    66176-005
  • O.D.×L:
    22×154 mm
  • Supplier No.:
    45290-154
Specifications
About this item
Used to determine the color of refined vegetable oils such as cotton seed, corn and soybean oils, as well as tallows, greases, fatty acids, etc., with wesson type tintometers. Also used to determine the color of whole and refined tall oils with a lovibond tintometer.
  • Shadowless bottom transmits undistorted light
  • Rings and legend are durable white ceramic enamel
  • Rings are located at specific distances from the inside bottom of the tube
  • For use with AOCS method Cc 13b
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.