ContactUsLinkComponent
Conical Bottom Autosampler Tubes with Graduation, PerkinElmer
Catalog # CA97024-502
Supplier:  PerkinElmer
undefined
Conical Bottom Autosampler Tubes with Graduation, PerkinElmer
Catalog # CA97024-502
Supplier:  PerkinElmer
Supplier Number:  B0193233
Specifications
  • Capacity:
    15 ml
  • Description:
    Standard Autosampler Tubes with Red Flat Caps Attached (Standard PerkinElmer S10 Tubes)
  • Size:
    17×120 mm
  • Cat. no.:
    CA97024-502
  • Packaging:
    50/bag
  • Supplier No.:
    B0193233
Specifications
About this item
These standard translucent polypropylene tubes are designed to provide excellent chemical resistance ensuring an accurate analysis.
  • Compatible with PerkinElmer Autosamplers for AA, ICP-OES and ICP-MS instruments
  • Tubes are graduated for ease-of-use
  • Writing patch area for sample identification
These are the standard autosampler tubes for the PerkinElmer S10 autosampler.