WHEATON® Testing Jars, DWK Life Sciences | Avantor
WHEATON® Testing Jars, DWK Life Sciences
WHEATON® Testing Jars, DWK Life Sciences
Certificates
WHEATON® Testing Jars, DWK Life Sciences
WHEATON® Testing Jars, DWK Life Sciences
2 Options Available Below
Product Details & Documents
Premium test jars with attached white polypropylene caps.
  • Made from USP Type III clear soda-lime glass
  • Wide mouth design is ideal for use with solids
  • Pre-attached white polypropylene caps with choice of liner
Walls allow for complete removal of contents. Ideal for soil sampling and environmental applications.
Product Family Options
Informations sur le produit
  • Capacité
    Diameter ×H
    Color
    Liner Material
    Availability
    Price
  • 60 ml (2 oz.)
    45×95 mm
    Clear
    Poly-vinyl
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 180
    Case of 180 - $737.10
    $737.10
    Prix promotionnel pour les nouveaux clients
    Case of 180
       $737.10
    Supplier Number: W216650
  • 60 ml (2 oz.)
    45×95 mm
    Clear
    PTFE faced foamed PE
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 180
    Case of 180 - $946.40
    $946.40
    Prix promotionnel pour les nouveaux clients
    Case of 180
       $946.40
    Supplier Number: W216651