KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences
KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences
Certificates
KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences
KIMBLE® MICROFLEX® Pear Shaped Flasks, Threaded, DWK Life Sciences
3 Options Available Below
Product Details & Documents
These flasks have short necks for easy retrieval of residues or reaction products.
  • Accessory for Microscale MICROFLEX® threaded kits
  • Thread size: 20-400
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Product Family Options
Informations sur le produit
  • Capacité
    Désignation
    Availability
    Price
  • 5 ml
    Pear-shaped flask, threaded neck, O.D×H: 25×50 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $32.58
    $32.58
    Prix promotionnel pour les nouveaux clients
    Each
       $32.58
    Supplier Number: 747540-0520
  • 10 ml
    Pear-shaped flask, threaded neck, O.D×H: 28×55 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $32.21
    $32.21
    Prix promotionnel pour les nouveaux clients
    Each
       $32.21
    Supplier Number: 747540-1020
  • 15 ml
    Pear-shaped flask, threaded neck, O.D×H: 30×55 mm
    Discontinued
    Supplier Number: -
  • 25 ml
    Pear-shaped flask, threaded neck, O.D×H: 80×39 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $45.41
    $45.41
    Prix promotionnel pour les nouveaux clients
    Each
       $45.41
    Supplier Number: 747540-2520