ContactUsLinkComponent
WHEATON® Diazomethane Generator, DWK Life Sciences
Catalog # 21518-134
Small Business Enterprise
undefined
WHEATON® Diazomethane Generator, DWK Life Sciences
Catalog # 21518-134
Supplier Number:  281155
Small Business Enterprise
Specifications
  • Description:
    Diazomethane generator, complete
  • Height:
    200 mm
  • Cat. no.:
    21518-134
  • Supplier No.:
    281155
Specifications
About this item
This Diazomethane Generator is used to generate diazomethane and other gases for gas chromatography, mass spectroscopy and NMR studies using one millimole of Methylnitronitrosoguanidine (MNNG).
  • Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to ASTM E 438 Type I, Class A requirements
Includes an “O” ring clamp, a 13-425 screw cap with a cut-out top and a PTFE faced rubber liner.
Caution: Please use a safety shield and efficient fume hood when generating diazomethane or similar gases.
O-rings
Product Information