BioClean™ Microflow Face Veil with Headloop, Nitritex | Avantor
BioClean™ Microflow Face Veil with Headloop, Nitritex
Supplier:   Ansell Canada
BioClean™ Microflow Face Veil with Headloop, Nitritex
Supplier:  Ansell Canada
Certificates
BioClean™ Microflow Face Veil with Headloop, Nitritex
BioClean™ Microflow Face Veil with Headloop, Nitritex
1 Options Available Below
Product Details & Documents
BioClean™ Microflow Cleanroom Face Veil is low-linting with ultrasonically sealed edges reducing the risk of contamination into the controlled environment.
  • Veil made from low linting thermabond non-woven material
  • White hydroentangled polyester binding and tubular polyester head loop
  • Processed in a cleanroom environment, contains no latex
Features a fully enclosed malleable aluminum nose-band with plastic coated and head-loops for a good fit. Suitable for use in class 10, ISO Class 4, M2.5 cleanrooms.
Certifications: Manufactured in a facility holding ISO 9001:2008.
Processed in an NEBB certified ISO class 4 cleanroom environment.
Sampling and inspection in accordance with ISO 2859-3:1994.
Packaging: 100 pieces per sealed inner PE bag; one inner bag per sealed outer PE bag; 10 outer bags per lined carton (1000/carton).
Product Family Options
Informations sur le produit
  • Color
    Sterility
    Largeur
    Longueur
    Availability
    Price
  • White
    Non sterile
    330 ±5 mm
    380 ±5 mm
    Discontinued
    Supplier Number: BFV05
  • White
    Non sterile
    330 ±5 mm
    380 ±5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,080.00
    $1,080.00
    Case of 1,000
       $1,080.00
    Supplier Number: BFV05