TechNitrile Class 100 TACKY Nitrile Gloves, TechNiGlove
TechNitrile Class 100 TACKY Nitrile Gloves, TechNiGlove
TechNitrile Class 100 TACKY Nitrile Gloves, TechNiGlove
Supplier:  Techniglove
Certificates
TechNitrile Class 100 TACKY Nitrile Gloves, TechNiGlove
TechNitrile Class 100 TACKY Nitrile Gloves, TechNiGlove
6 Options Available Below
Product Details & Documents
TechniCleaned TACKY ambidextrous 12" Class 100 nitrile gloves have a tacky surface to enhance grip.
  • TACKY surface for enhanced grip
  • Class 100
  • 100% nitrile
  • Ambidextrous
  • Powder-free
  • Batch specific COC available
TechNiGlove products are 100% nitrile, contain no natural rubber or silicone, and meet the most stringent requirements for particles and extractables. New soft modulus nitrile delivers the fit and feel of latex for the ultimate stretch and comfort. Microtextured fingertips provide an excellent grip on all surfaces.
Suitable for use in cleanrooms and contamination controlled work environments, pharmaceutical, medical device, disc drive, and semiconductor manufacturing.
Packaging: Packaged 100 gloves per double poly-sealed bag, 10 bags per case.
Specifications for Product Family
Specifications
  • Catalog No:
  • 76378-002
  • 76378-004
  • 76378-006
  • 76378-008
  • 76378-010
  • 76378-012
  • Ambidextrous/Hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Powdered/Powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Microtextured fingertips
  • Microtextured fingertips
  • Microtextured fingertips
  • Microtextured fingertips
  • Microtextured fingertips
  • Microtextured fingertips
Specifications
Product Family Options
Product Information
  • Taille du gant
    Glove Color
    Longueur
    Épaisseur
    Availability
    Price
  • XS
    White
    30.5 cm (12")
    5 mil.
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $696.30
    $696.30
    New Customer Promotional Price
    Case of 1,000
       $696.30
    Supplier Number: TN1000WT
    Specifications:
    More
    Specifications
    Cat. No.
    76378-002
    Ambidextrous/Hand-specific
    Ambidextrous
    Powdered/Powder-free
    Powder-free
    Finish
    Microtextured fingertips
  • S
    White
    30.5 cm (12")
    5 mil.
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $777.49
    $777.49
    New Customer Promotional Price
    Case of 1,000
       $777.49
    Supplier Number: TN1001WT
    Specifications:
    More
    Specifications
    Cat. No.
    76378-004
    Ambidextrous/Hand-specific
    Ambidextrous
    Powdered/Powder-free
    Powder-free
    Finish
    Microtextured fingertips
  • M
    White
    30.5 cm (12")
    5 mil.
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $779.61
    $779.61
    New Customer Promotional Price
    Case of 1,000
       $779.61
    Supplier Number: TN1002WT
    Specifications:
    More
    Specifications
    Cat. No.
    76378-006
    Ambidextrous/Hand-specific
    Ambidextrous
    Powdered/Powder-free
    Powder-free
    Finish
    Microtextured fingertips
  • L
    White
    30.5 cm (12")
    5 mil.
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $723.08
    $723.08
    New Customer Promotional Price
    Case of 1,000
       $723.08
    Supplier Number: TN1003WT
    Specifications:
    More
    Specifications
    Cat. No.
    76378-008
    Ambidextrous/Hand-specific
    Ambidextrous
    Powdered/Powder-free
    Powder-free
    Finish
    Microtextured fingertips
  • XL
    White
    30.5 cm (12")
    5 mil.
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $592.44
    $592.44
    New Customer Promotional Price
    Case of 1,000
       $592.44
    Supplier Number: TN1004WT
    Specifications:
    More
    Specifications
    Cat. No.
    76378-010
    Ambidextrous/Hand-specific
    Ambidextrous
    Powdered/Powder-free
    Powder-free
    Finish
    Microtextured fingertips
  • XXL
    White
    30.5 cm (12")
    5 mil.
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $690.41
    $690.41
    New Customer Promotional Price
    Case of 1,000
       $690.41
    Supplier Number: TN1005WT
    Specifications:
    More
    Specifications
    Cat. No.
    76378-012
    Ambidextrous/Hand-specific
    Ambidextrous
    Powdered/Powder-free
    Powder-free
    Finish
    Microtextured fingertips