Thermometer Centering Device, DWK Life Sciences
Thermometer centering device
29618-006
: 26015C-125
Designed for 6.5 mm O.D. manual thermometer or 1/4" temperature sensor probe as used on standard automated distillation apparatus with flasks ( 26015-125).
- Fits glassware designed for rubber stopper size 2
- PTFE body with FKM O-ring seal assures proper centering of sensor probe in flask neck
- Cap material is yellow polypropylene
: ASTM Method D86.
- Désignation:Thermometer centering device