MIDI-VAP 3000 Combination Distillation Systems Components, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
À propos de cet article
These components are used for Midi-Vap 3000 distillation systems which are designed to support the semiautomatic colorimetry methods for drinking, ground, surface and saline waters - as well as domestic and industrial wastes.
- Uses a higher, adjustable, temperature range
- Thermostatically controlled to avoid overheating
- Ammonia and phenols can be run simultaneously
Certifications: UL / CSA certified.
Caution: Midi-phenol and Midi-ammonia reactions produce toxic HCN gas. Perform these reactions under a well-ventilated fume hood.
Frequently Bought Together