Overpack Poly Drums, Eagle Mfg | Avantor
Overpack Poly Drums, Eagle Mfg
Overpack with screw-on lid, 65 gal., 31 O.Ø×33.75" H, Yellow
Catalog # 76369-708
Supplier:   Eagle Manufacturing
Fournisseur #:  1661
Overpack Poly Drums, Eagle Mfg
Catalog # 76369-708
Overpack Poly Drums, Eagle Mfg
Overpack Poly Drums, Eagle Mfg
Overpack with screw-on lid, 65 gal., 31 O.Ø×33.75" H, Yellow
Price
$601.17
Log in pour la disponibilité
Each
Product Details & Documents
These overpack drums are constructed of blow-molded HDPE and are durable and lightweight.
  • Chemical and weather resistant
  • UV protection
  • Approved for use with packing groups i, ii and iii
Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration. For use with larger non damaged containers, cans and drums of non leaking hazardous materials.
Certifications: UN certified. UL rating. Food grade quality - FDA 21 CFR 177.1520.
Spécifications
  • Capacité:
    65 gal.
  • Color:
    Yellow
  • Désignation:
    Overpack with screw-on lid
  • Dimensions:
    31 O.Ø×33.75" H