Lab Pack Poly Drums, Eagle Mfg
Lab Pack Poly Drums, Eagle Mfg
Supplier:   Eagle Manufacturing
Lab Pack Poly Drums, Eagle Mfg
Certificates
Lab Pack Poly Drums, Eagle Mfg
Lab Pack Poly Drums, Eagle Mfg
18 Options Available Below
Product Details & Documents
These lab pack drums are constructed of blow-molded HDPE and are durable and lightweight.
  • Chemical and weather resistant
  • UV protection
  • Approved for use with packing groups i, ii and iii
Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration. For use with smaller non-damaged bottles, cans, carboys and 5-gallon pails of non-leaking hazardous materials.
Certifications: UN certified. UL rating. Food grade quality - FDA 21 CFR 177.1520.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Weight
    Dimensions
    Color
    Lid Type
    UN/DOT Rating
    Availability
    Price
  • Lab pack poly drum, with screw-on lid
    30 gal.
    18 lbs.
    19.88" (50.5 cm) top×16.25" (41.3 cm) bottom, 28.25" (71.8 cm) height
    Yellow
    Regular
    X120/Y180
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $223.52
    $223.52
    Each
       $223.52
    Supplier Number: 1600SL
  • Lab pack open head plastic barrel drum with 1×2 and 1x¾" bung holes, metal lever-lock
    30 gal.
    18 lbs.
    19 (48.5cm) top, 16.25" (41.3cm) bottom, 26.38" (67cm) height"
    Blue
    Bung
    1H2/X120/S, 1H2/Y180/S
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $141.88
    $141.88
    Each
       $141.88
    Supplier Number: 1601MBBG
  • Lab pack poly drum, open head, with metal lever-lock ring, 16 gauge metal lever band
    30 gal.
    18 lbs.
    19" (48.5 cm) top, 16.25" (41.3 cm) bottom, 26.38" (67 cm) height
    Blue
    Regular
    1H2/X120/S, 1H2/Y180/S
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $235.71
    $235.71
    Each
       $235.71
    Supplier Number: 1601MB
  • Lab pack open head plastic barrel drum with 1×2 and 1x¾" bung holes, metal lever-lock, 16 gauge metal lever band
    30 gal.
    18 lbs.
    19 (48.5cm) top, 16.25" (41.3cm) bottom, 26.38" (67cm) height"
    Black
    Bung
    1H2/X120/S, 1H2/Y180/S
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $135.69
    $135.69
    Each
       $135.69
    Minimum order quantity is 1
    Supplier Number: 1601MBLKBG
  • Lab pack open head plastic barrel drum with 1×2 and 1x¾" bung holes, plastic lever-lock
    30 gal.
    18 lbs.
    19 (48.5cm) top, 16.25" (41.3cm) bottom, 26.38" (67cm) height"
    Black
    Bung
    1H2/X100/S, 1H2/Y180/S
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $130.68
    $130.68
    Each
       $130.68
    Minimum order quantity is 1
    Supplier Number: 1601BLKBG
  • Lab pack open head plastic barrel drum with 2×2 bung hole, metal bolt ring
    30 gal.
    18 lbs.
    19 (48.5cm) top, 16.25" (41.3cm) bottom, 26.38" (67cm) height"
    Black
    Bung
    1H2/X120/S, 1H2/Y180/S
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $178.76
    $178.76
    Each
       $178.76
    Minimum order quantity is 1
    Supplier Number: 1601MBK-BRBG2
  • Lab pack open head plastic barrel drum with 2×2 bung holes, metal bolt ring
    30 gal.
    18 lbs.
    19 (48.5cm) top, 16.25" (41.3cm) bottom, 26.38" (67cm) height"
    Yellow
    Bung
    1H2/X120/S, 1H2/Y180/S
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $189.62
    $189.62
    Each
       $189.62
    Minimum order quantity is 1
    Supplier Number: 1601M-BRBG2
  • Lab pack open head plastic barrel drum with 2×2 bung holes, metal bolt rings
    30 gal.
    18 lbs.
    19 (48.5cm) top, 16.25" (41.3cm) bottom, 26.38" (67cm) height"
    Blue
    Bung
    1H2/X120/S, 1H2/Y180/S
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $189.52
    $189.52
    Each
       $189.52
    Minimum order quantity is 1
    Supplier Number: 1601MB-BRBG2
  • Lab pack open head plastic barrel drum with 2×2 bung holes, metal lever-lock
    30 gal.
    18 lbs.
    19 (48.5cm) top, 16.25" (41.3cm) bottom, 26.38" (67cm) height"
    Yellow
    Bung
    1H2/X120/S, 1H2/Y180/S
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $183.91
    $183.91
    Each
       $183.91
    Minimum order quantity is 1
    Supplier Number: 1601M-BG2
  • Lab pack open head plastic barrel drum, metal lever-lock, plastic lever with down leg
    30 gal.
    18 lbs.
    19" (48.5cm) Top, 16.25" (41.3cm) Bottom, 26.38" (67cm) Height
    Yellow
    Regular
    1H2/X120/S, 1H2/Y180/S
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $128.27
    $128.27
    Each
       $128.27
    Supplier Number: 1601
  • Lab pack open head plastic barrel drum, metal lever-lock, 16 gauge metal lever band
    30 gal.
    18 lbs.
    19" (48.5cm) Top, 16.25" (41.3cm) Bottom, 26.38" (67cm) Height
    Yellow
    Regular
    1H2/X120/S, 1H2/Y180/S
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $139.22
    $139.22
    Each
       $139.22
    Supplier Number: 1601M
  • Lab pack open head plastic barrel drum, plastic lever-lock
    30 gal.
    16 lbs.
    19" (48.5cm) Top, 16.25" (41.3cm) Bottom, 26.38" (67cm) Height
    Blue
    Regular
    1H2/X100/S, 1H2/Y180/S
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $139.55
    $139.55
    Each
       $139.55
    Supplier Number: 1601B
  • Lab pack plastic barrel drum, plastic lever-lock
    30 gal.
    18 lbs.
    19" (48.5cm) Top, 16.25" (41.3cm) Bottom, 26.38" (67cm) Height
    Black
    Regular
    1H2/X100/S, 1H2/Y180/S
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $126.86
    $126.86
    Each
       $126.86
    Supplier Number: 1601BLK
  • Lab pack open head plastic barrel drum, plastic lever-lock
    30 gal.
    16 lbs.
    19" (48.5cm) Top, 16.25" (41.3cm) Bottom, 26.38" (67cm) Height
    Red
    Regular
    1H2/X100/S, 1H2/Y180/S
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $140.12
    $140.12
    Each
       $140.12
    Supplier Number: 1601RED
  • Lab pack plastic barrel drum, black, metal lever-lock
    30 gal.
    18 lbs.
    19" (48.5cm) Top, 16.25" (41.3cm) Bottom, 26.38" (67cm) Height
    Black
    Regular
    1H2/X120/S, 1H2/Y180/S
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $134.46
    $134.46
    Each
       $134.46
    Minimum order quantity is 1
    Supplier Number: 1601MBLK
  • Lab pack open head plastic barrel drum, metal bolt ring
    30 gal.
    18 lbs.
    19" (48.5cm) Top, 16.25" (41.3cm) Bottom, 26.38" (67cm) Height
    Blue
    Regular
    1H2/X120/S, 1H2/Y180/S
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $129.35
    $129.35
    Each
       $129.35
    Supplier Number: 1601MBBR
  • Lab pack open head plastic barrel drum with 2×2 bung holes, metal lever-lock
    30 gal.
    18 lbs.
    19" (48.5cm) Top, 16.25" (41.3cm) Bottom, 26.38" (67cm) Height
    Blue
    Bung
    1H2/X120/S, 1H2/Y180/S
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $183.91
    $183.91
    Each
       $183.91
    Minimum order quantity is 1
    Supplier Number: 1601MB-BG2
  • Lab pack open head plastic barrel drum with 2×2 bung holes, metal lever-lock
    30 gal.
    18 lbs.
    19" (48.5cm) Top, 16.25" (41.3cm) Bottom, 26.38" (67cm) Height
    Black
    Bung
    1H2/X120/S, 1H2/Y180/S
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $173.29
    $173.29
    Each
       $173.29
    Minimum order quantity is 1
    Supplier Number: 1601MBK-BG2