KIMBLE® MICROFLEX™ Long Neck Threaded Boiling Flasks, DWK Life Sciences | Avantor
KIMBLE® MICROFLEX™ Long Neck Threaded Boiling Flasks, DWK Life Sciences
KIMBLE® MICROFLEX™ Long Neck Threaded Boiling Flasks, DWK Life Sciences
Certificates
KIMBLE® MICROFLEX™ Long Neck Threaded Boiling Flasks, DWK Life Sciences
KIMBLE® MICROFLEX™ Long Neck Threaded Boiling Flasks, DWK Life Sciences
2 Options Available Below
Product Details & Documents
These long neck threaded boiling flasks are used as an accessory for Microscale MICROFLEX™ threaded kits.
  • Manufactured from 33 expansion, low extractable borosilicate glass
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Product Family Options
Informations sur le produit
  • Capacité
    Désignation
    Hauteur
    Ø ext.
    Availability
    Price
  • 10 ml
    Long neck threaded round bottom boiling flasks, thread size 20-400
    60 mm
    31 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $32.58
    $32.58
    Prix promotionnel pour les nouveaux clients
    Each
       $32.58
    Supplier Number: 747520-1020
  • 25 ml
    Long neck threaded round bottom boiling flasks, thread size 20-400
    65 mm
    42 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $39.04
    $39.04
    Prix promotionnel pour les nouveaux clients
    Each
       $39.04
    Supplier Number: 747520-2520