WHEATON® Pasteur Pipette Adapter, DWK Life Sciences
Adapter, 2 ml
Supplier: DWK Life Sciences (KIMBLE)
Fournisseur #: W820125
WHEATON® Pasteur Pipette Adapter, DWK Life Sciences
Catalog # 75800-372
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
Product Details & Documents
PVDF adapter fits pipette nozzle to accommodate standard glass Pasteur pipettes for model 832 and 835.
The adapter has two FPM O-rings guaranteeing proper holding and tightness of standard glass 2 ml Pasteur pipettes.
Spécifications
- Désignation:Adapter, 2 ml