WHEATON® Tablet Square Bottles, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
High-quality tablet square bottles are made from USP Type III soda-lime glass.
- Wide mouth design is ideal for use with solids
- Pre-attached white polypropylene caps with PE cone liner
Premium bottles come with attached screw caps.
Frequently Bought Together