KIMBLE® ULTRA-WARE® Conversion Base, DWK Life Sciences | Avantor
KIMBLE® ULTRA-WARE® Conversion Base, DWK Life Sciences
ULTRA-WARE® conversion base, 90 mm
Catalog # KT953984-0090
Fournisseur #:  953984-0090
KIMBLE® ULTRA-WARE® Conversion Base, DWK Life Sciences
Catalog # KT953984-0090
KIMBLE® ULTRA-WARE® Conversion Base, DWK Life Sciences
KIMBLE® ULTRA-WARE® Conversion Base, DWK Life Sciences
ULTRA-WARE® conversion base, 90 mm
Price
$1,232.50
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
ULTRA-WARE® conversion base converts the five valve, four valve and filtration caps to use 90 mm filter membranes, providing up to four times faster filtration.
  • Requires a 90 mm solvent pickup adapter
The conversion base is manufactured from glass-filled PTFE and is supplied with a coarse porosity (40 to 60 micron porosity) glass support frit and an upper screw clamp.
Spécifications
  • Désignation:
    ULTRA-WARE® conversion base
  • Taille:
    90 mm
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Solvent Pickup Adapter, THF Resistant, 9.0 cm
    Solvent Pickup Adapter, THF Resistant, 9.0 cm
    KT953906-6390
    Log in pour la disponibilité
    Each - $1,101.10
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Solvent Pickup Adapter, 9.0 cm
    Solvent Pickup Adapter, 9.0 cm
    KT953906-0090
    Log in pour la disponibilité
    Each - $716.55
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Glass support frit, 90 mm, 40 - 60 micron porosity
    Glass support frit, 90 mm, 40 - 60 micron porosity
    KT953914-0091
    Log in pour la disponibilité
    Each - $353.99
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Upper screw clamp, 90 mm
    Upper screw clamp, 90 mm
    KT953914-0092
    Log in pour la disponibilité
    Each - $271.18