Polytron® Generator, PT-DA/2WEC-E192, Kinematica
Supplier: Kinematica
Jump to:
About this item
W-design for fibrous/stringy materials. Special design to prevent the teeth clogging. Dispersing generators are made from high-alloy stainless steel 316L, electropolished, Ra≤1.6 μm as standard.
- Dispersing fibrous, stringy or solid samples (e.g., pieces of meat)
- Standard dispersing, emulsifying and suspending tasks
3 Options Available Below
Product Details & Documents
W-design for fibrous/stringy materials. Special design to prevent the teeth clogging. Dispersing generators are made from high-alloy stainless steel 316L, electropolished, Ra≤1.6 μm as standard.
- Dispersing fibrous, stringy or solid samples (e.g., pieces of meat)
- Standard dispersing, emulsifying and suspending tasks
Product Family Options
Product Information
- DésignationSample VolumeDiameter ×LFor Use WithAvailabilityPrice
- Polytron® 20 mm generator, PT-DA 20/2WEC-E19210 - 2500 ml20×192 mmPT 2500 ELog in for availabilityEach$5,599.47Supplier Number: 11030258
- Polytron® 20 mm generator, PT-DA 20/2WEC-E19210 - 2500 ml20×192 mmPT 2500 ELog in for availabilityEach$5,929.41Supplier Number: 11030276
- Polytron® 25 mm generator, PT-DA 25/2WEC-E192100 - 2500 ml25×192 mmPT 2500 ELog in for availabilityEach$6,381.74Supplier Number: 11030260