Polytron® Generator, PT-DA/BEC (BIOTRONA), Kinematica
Polytron® Generator, PT-DA/BEC (BIOTRONA), Kinematica
Supplier:   Kinematica
Polytron® Generator, PT-DA/BEC (BIOTRONA), Kinematica
Supplier:  Kinematica
Certificates
Polytron® Generator,  PT-DA/BEC (BIOTRONA), Kinematica
Polytron® Generator, PT-DA/BEC (BIOTRONA), Kinematica
4 Options Available Below
Product Details & Documents
BIOTRONA® design high turbulence mixing. High-turbulence mixing head with low shear forces and energy inputs. Time saving with improved results compared to stirrers and dissolvers of up to 90%. Dispersing generators are made from high-alloy stainless steel 316L, electropolished, Ra≤1.6 μm as standard.
  • Fast dissolving and suspending of solid particles into liquids
  • Ideal for higher viscosities
  • Lower shear forces
Product Family Options
Product Information
  • Désignation
    Sample Volume
    Diameter ×L
    For Use With
    Availability
    Price
  • Polytron® 7 mm generator, PT-DA 07/BEC-E107 (BIOTRONA) 
    10 - 100 ml
    7×107 mm
    PT 1200 E, PT 1300 D, PT 2500 E
    Log in for availability
    Log in for availability
    Each
    Each - $4,699.29
    $4,699.29
    Each
       $4,699.29
    Minimum order quantity is 1
    Supplier Number: 11030034
  • Polytron® 12 mm generator, PT-DA 12/BEC-E157 (BIOTRONA)
    50 - 500 ml
    12×157 mm
    PT 1200 E, PT 1300 D, PT 2500 E
    Log in for availability
    Log in for availability
    Each
    Each - $5,381.54
    $5,381.54
    Each
       $5,381.54
    Supplier Number: 11030286
  • Polytron® 20 mm generator, PT-DA 20/BEC-E192 (BIOTRONA)
    100 - 2000 ml
    20×192 mm
    PT 2500 E
    Log in for availability
    Log in for availability
    Each
    Each - $4,986.19
    $4,986.19
    Each
       $4,986.19
    Minimum order quantity is 1
    Supplier Number: 11030288
  • Polytron® 25 mm generator, PT-DA 25/BEC-E192 (BIOTRONA)
    100 - 2500 ml
    25×192 mm
    PT 2500 E
    Log in for availability
    Log in for availability
    Each
    Each - $6,071.16
    $6,071.16
    Each
       $6,071.16
    Supplier Number: 11030291