Polytron® Generator, PT-DA/2EC-E192, Kinematica | Avantor
Polytron® Generator, PT-DA/2EC-E192, Kinematica
Supplier:   Kinematica
Polytron® Generator, PT-DA/2EC-E192, Kinematica
Supplier:  Kinematica
Certificates
Polytron® Generator, PT-DA/2EC-E192, Kinematica
Polytron® Generator, PT-DA/2EC-E192, Kinematica
2 Options Available Below
Product Details & Documents
Standard dispersing generators come with the EC (EasyClean) design and rotor/stator geometries with two, four or six teeth rows. This is all you need for popular applications in the preparation of samples, formulation development and small-scale production. Dispersing generators are made from high-alloy stainless steel 316L, electropolished, Ra≤1.6 μm as standard.
  • All standard dispersing, emulsifying and suspending tasks
  • Dispersing of plant, human or animal tissues, resins and pigments in a variety of fluids and volumes
  • Sample preparation for extraction and dissolution of organic materials
  • Intensive mixing, dissolving solids
Product Family Options
Informations sur le produit
  • Désignation
    Sample Volume
    Diameter ×L
    For Use With
    Availability
    Price
  • Polytron® 20 mm generator, PT-DA 20/2EC-E192
    100 - 2000 ml
    20×192 mm
    PT 2500 E
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,619.75
    $4,619.75
    Each
       $4,619.75
    Supplier Number: 11030069
  • Polytron® 20 mm generator, PT-DA 25/2EC-E192
    100 - 2500 ml
    25×192 mm
    PT 2500 E
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,713.08
    $5,713.08
    Each
       $5,713.08
    Supplier Number: 11030264