KIMBLE® MICROFLEX® Erlenmeyer Flasks, Threaded, DWK Life Sciences
KIMBLE® MICROFLEX® Erlenmeyer Flasks, Threaded, DWK Life Sciences
KIMBLE® MICROFLEX® Erlenmeyer Flasks, Threaded, DWK Life Sciences
Certificates
KIMBLE® MICROFLEX® Erlenmeyer Flasks, Threaded, DWK Life Sciences
KIMBLE® MICROFLEX® Erlenmeyer Flasks, Threaded, DWK Life Sciences
2 Options Available Below
Product Details & Documents
These threaded Erlenmeyer flasks are useful for sample collection, derivatization reactions, extractions, sample storage and micro mixing.
  • Without graduations
  • Manufactured from 33 expansion, low extractable borosilicate glass
Accessory for Microscale MICROFLEX™ threaded kits.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Documents
Product Family Options
Product Information
  • Désignation
    Capacité
    Hauteur
    Epaisseur
    Availability
    Price
  • MICROFLEX® threaded erlenmeyer flasks
    10 ml
    45 mm
    20 - 400
    Log in for availability
    Log in for availability
    Each
    Each - $42.41
    $42.41
    New Customer Promotional Price
    Each
       $42.41
    Supplier Number: 747530-1020
  • MICROFLEX® threaded erlenmeyer flasks
    25 ml
    60 mm
    20 - 400
    Log in for availability
    Log in for availability
    Each
    Each - $39.95
    $39.95
    New Customer Promotional Price
    Each
       $39.95
    Supplier Number: 747530-2520