Wizard Series PVC/Nylon/PVC 60" Coats with Detachable Hood, MCR Safety
This wizard series coat features a 0.28 mm double coating of PVC, with tough nylon scrim material layered in between.
- 60" ankle length design
- Detachable drawstring hood
- Snaps on front and back can fit together to form chaps
- Rip stop scrim material provides excellent resistance to snags and tears
Nylon scrim has excellent rip and puncture resistance. This yellow two-piece coat is a 60" (ankle length) option that features a detachable drawstring hood and storm flap under a snap up front closure. Snaps on the lower front and back can fit together to form 'chaps' around each leg. Limited flammability, this garment is not intended for use as protection against ARC flash or flash fire.
- Catalog No:
- 76341-308
- 76341-844
- 76341-862
- 76341-696
- 76341-520
- 76341-782
- 76341-430
- 76341-818
- 76341-620
- Garment application:
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- PVC/Nylon/PVC
- Thickness:
- 0.28 mm
- 0.28 mm
- 0.28 mm
- 0.28 mm
- 0.28 mm
- 0.28 mm
- 0.28 mm
- 0.28 mm
- 0.28 mm
Frequently Bought Together