Rake Retractor
Rake retractor, 19.1 cm (7½"), Stainless steel
Catalog # 470321-010
Supplier: United Scientific Supplies
: RRET06
Jump to:
Product Details & Documents
Easily manipulate heavier tissue.
- Wide handle For easy manipulation
- Three teeth provide a secure grip
- Long handle provides space for students working in teams
Used for tissue manipulation during dissection, the rake retractor has three recurved teeth to pull tissue away from structures. Use of the retractor by an assistant, allows a student to make very clean and precise cuts separating the tissue from an underlying structure.
Spécifications
- Désignation:Rake retractor
- Longueur:19.1 cm (7½")
- Matière:Stainless steel