Human Skeleton Model with Fold-Out Guide
Human skeleton model with fold-out guide
Catalog # 470320-998
Supplier: United Scientific Supplies
Fournisseur #: MASKE1
Jump to:
Product Details & Documents
Color coded Skeleton with comprehensive manual.
- Foldout guide rests in a slot molded into the plastic base
Skeleton model is molded from PVC plastic and hand-painted to show structures of the axial skeleton. Skeleton measures 32×8× 5" and mounts on a metal rod with a plastic stand.
Axial skeleton parts, comprised of the skull, ossicles, hyoid bone, rib cage, sternum and vertebral column, are painted blue. Includes a fold-out laminated color guide indicating 143 labeled structures including anterior/posterior views of the skeleton and detail of spine, pelvis, vertebra, joint, rib cage, hand and foot. One side of the guide includes structure names. The other side includes only numbers so it can be used for student assessment. Also includes a 12-page reproducible study guide.
Spécifications
- Désignation:Human skeleton model with fold-out guide