Changes in State Stem Kit | Avantor
Changes in State Stem Kit
Changes in state stem kit
Catalog # 470320-952
Fournisseur #:  AISCSKIT
Changes in State Stem Kit
Catalog # 470320-952
Changes in State Stem Kit
Changes in State Stem Kit
Changes in state stem kit
Price
$449.50
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Two investigative areas covering six STEM activities.
  • Complete classroom solution
  • Activity manual included
This kit teaches students about heats of fusion and reaction, thermochemistry, iceberg types and thermohaline circulation and climate change.
 : The kit includes enough materials for 40 students working in groups of four, as well as PDF teacher and student guides and other digital content.
Spécifications
  • Désignation:
    Changes in state stem kit