Embedding Blocks
À propos de cet article
Embedding blocks for biological specimens.
- Wide range of sizes
- Made of phenolic resin
Used for celloidin or paraffin sections, these embedding blocks are made of phenolic resin, which can be marked with a glass marking pencil. Top surface has 1/32" deep grid to provide a firm hold for embedding specimen. Blocks are ¾" high. Sold in packs of 10.
4 Options Available Below
Product Details & Documents
Embedding blocks for biological specimens.
- Wide range of sizes
- Made of phenolic resin
Used for celloidin or paraffin sections, these embedding blocks are made of phenolic resin, which can be marked with a glass marking pencil. Top surface has 1/32" deep grid to provide a firm hold for embedding specimen. Blocks are ¾" high. Sold in packs of 10.
Product Family Options
Informations sur le produit
- DésignationDimensionsAvailabilityPrice
- Supplier Number: EB0001
- Supplier Number: EB0002
- Supplier Number: EB0003
- Supplier Number: EB0004